Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39307657-39307898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN1
Entrez Gene ID
57622
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Gene Aliases
SALM2
UniGene
Hs.97860
Gene Chromosome Location
Chr.19: 39306817-39320867 on Build GRCh38
Transcripts
NM_020862.1 XM_005259102.4 XM_017027033.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
711511 c.1998C>A p.F666L Chr.19: 39307807-39307807 on Build GRCh38
6702825 c.1940C>T p.A647V Chr.19: 39307865-39307865 on Build GRCh38
5157497 c.1950G>T p.P650P Chr.19: 39307855-39307855 on Build GRCh38
4477603 c.2024C>T p.A675V Chr.19: 39307781-39307781 on Build GRCh38
4698673 c.1978G>A p.G660R Chr.19: 39307827-39307827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771479196 Chr.19: 39307856-39307856 on Build GRCh38
rs540610015 Chr.19: 39307681-39307681 on Build GRCh38
rs780955035 Chr.19: 39307827-39307827 on Build GRCh38
rs62643365 Chr.19: 39307724-39307724 on Build GRCh38
rs368093101 Chr.19: 39307747-39307747 on Build GRCh38
rs772487093 Chr.19: 39307709-39307709 on Build GRCh38
rs910791388 Chr.19: 39307872-39307872 on Build GRCh38
rs1013179849 Chr.19: 39307707-39307707 on Build GRCh38
rs942202957 Chr.19: 39307873-39307873 on Build GRCh38
rs201713211 Chr.19: 39307728-39307728 on Build GRCh38
rs751453455 Chr.19: 39307792-39307792 on Build GRCh38
rs759817303 Chr.19: 39307751-39307751 on Build GRCh38
rs375012875 Chr.19: 39307831-39307831 on Build GRCh38
rs779717187 Chr.19: 39307834-39307834 on Build GRCh38
rs370561531 Chr.19: 39307785-39307785 on Build GRCh38
rs761258075 Chr.19: 39307843-39307843 on Build GRCh38
rs773598933 Chr.19: 39307842-39307842 on Build GRCh38
rs774844545 Chr.19: 39307700-39307700 on Build GRCh38
rs1032403117 Chr.19: 39307756-39307756 on Build GRCh38
rs769355365 Chr.19: 39307675-39307675 on Build GRCh38
rs527402445 Chr.19: 39307759-39307759 on Build GRCh38
rs765455703 Chr.19: 39307753-39307753 on Build GRCh38
rs757048407 Chr.19: 39307819-39307819 on Build GRCh38
rs768156325 Chr.19: 39307841-39307841 on Build GRCh38
rs759877931 Chr.19: 39307865-39307865 on Build GRCh38
rs988162501 Chr.19: 39307836-39307836 on Build GRCh38
rs35917023 Chr.19: 39307823-39307824 on Build GRCh38
rs758460743 Chr.19: 39307775-39307775 on Build GRCh38
rs765583425 Chr.19: 39307867-39307867 on Build GRCh38
rs954727334 Chr.19: 39307866-39307866 on Build GRCh38
rs762427646 Chr.19: 39307705-39307705 on Build GRCh38
rs752912824 Chr.19: 39307871-39307871 on Build GRCh38
rs1030167845 Chr.19: 39307850-39307850 on Build GRCh38
rs745572870 Chr.19: 39307829-39307829 on Build GRCh38
rs777113799 Chr.19: 39307861-39307861 on Build GRCh38
rs766784830 Chr.19: 39307745-39307745 on Build GRCh38

View Full Product Details