Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 65579427-65579652 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPWD1
Entrez Gene ID
23398
Gene Name
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases
-
UniGene
Hs.121432
Gene Chromosome Location
Chr.5: 65563236-65587549 on Build GRCh38
Transcripts
NM_001278926.1 NM_001278927.1 NM_001278929.1 NM_015342.3 XM_005248474.3 XM_006714583.3 XM_011543295.1 XM_017009286.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5918792 c.1297C>T p.Q433* Chr.5: 65579560-65579560 on Build GRCh38
1738193 c.1289A>G p.Q430R Chr.5: 65579552-65579552 on Build GRCh38
1738194 c.1293T>G p.N431K Chr.5: 65579556-65579556 on Build GRCh38
4717331 c.1282delG p.V428fs*11 Chr.5: 65579545-65579545 on Build GRCh38
1069352 c.1259A>T p.E420V Chr.5: 65579522-65579522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368870571 Chr.5: 65579544-65579544 on Build GRCh38
rs147425015 Chr.5: 65579575-65579575 on Build GRCh38
rs762575439 Chr.5: 65579503-65579503 on Build GRCh38
rs771883335 Chr.5: 65579464-65579464 on Build GRCh38
rs750458713 Chr.5: 65579519-65579519 on Build GRCh38
rs142815878 Chr.5: 65579504-65579504 on Build GRCh38
rs534130628 Chr.5: 65579602-65579602 on Build GRCh38
rs780334568 Chr.5: 65579535-65579535 on Build GRCh38
rs992267072 Chr.5: 65579543-65579543 on Build GRCh38
rs756254614 Chr.5: 65579526-65579526 on Build GRCh38
rs759061607 Chr.5: 65579483-65579483 on Build GRCh38
rs768800520 Chr.5: 65579545-65579545 on Build GRCh38
rs566310669 Chr.5: 65579553-65579553 on Build GRCh38
rs762085652 Chr.5: 65579610-65579610 on Build GRCh38
rs769280084 Chr.5: 65579577-65579577 on Build GRCh38
rs961101722 Chr.5: 65579539-65579539 on Build GRCh38
rs751913002 Chr.5: 65579508-65579508 on Build GRCh38
rs774916161 Chr.5: 65579586-65579586 on Build GRCh38
rs752343142 Chr.5: 65579491-65579491 on Build GRCh38
rs561055896 Chr.5: 65579562-65579562 on Build GRCh38
rs200778132 Chr.5: 65579518-65579518 on Build GRCh38
rs201676226 Chr.5: 65579598-65579598 on Build GRCh38
rs768057682 Chr.5: 65579616-65579616 on Build GRCh38
rs764805341 Chr.5: 65579485-65579485 on Build GRCh38
rs760629470 Chr.5: 65579470-65579470 on Build GRCh38
rs772928163 Chr.5: 65579469-65579469 on Build GRCh38
rs114773980 Chr.5: 65579456-65579456 on Build GRCh38
rs538651392 Chr.5: 65579509-65579509 on Build GRCh38
rs770803708 Chr.5: 65579480-65579480 on Build GRCh38
rs762638542 Chr.5: 65579589-65579589 on Build GRCh38

View Full Product Details