Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 156329643-156329898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR4A2
Entrez Gene ID
4929
Gene Name
nuclear receptor subfamily 4 group A member 2
Gene Aliases
HZF-3, NOT, NURR1, RNR1, TINUR
UniGene
Hs.563344
Gene Chromosome Location
Chr.2: 156324432-156334683 on Build GRCh38
Transcripts
NM_006186.3 XM_005246621.3 XM_005246622.3 XM_006712553.3 XM_011511246.1 XM_017004219.1 XM_017004220.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5769972 c.537G>A p.T179T Chr.2: 156329683-156329683 on Build GRCh38
162986 c.299A>G p.Y100C Chr.2: 156329888-156329888 on Build GRCh38
5228775 c.430delG p.D144fs*19 Chr.2: 156329757-156329757 on Build GRCh38
6729299 c.470C>T p.P157L Chr.2: 156329750-156329750 on Build GRCh38
3568135 c.528C>T p.F176F Chr.2: 156329659-156329659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201960934 Chr.2: 156329788-156329788 on Build GRCh38
rs141187633 Chr.2: 156329824-156329824 on Build GRCh38
rs377674598 Chr.2: 156329677-156329677 on Build GRCh38
rs374632208 Chr.2: 156329751-156329751 on Build GRCh38
rs773777485 Chr.2: 156329866-156329866 on Build GRCh38
rs746699693 Chr.2: 156329833-156329833 on Build GRCh38
rs746994082 Chr.2: 156329733-156329733 on Build GRCh38
rs775971579 Chr.2: 156329734-156329734 on Build GRCh38
rs569665455 Chr.2: 156329695-156329695 on Build GRCh38
rs774629025 Chr.2: 156329861-156329862 on Build GRCh38
rs778802748 Chr.2: 156329708-156329708 on Build GRCh38
rs781377163 Chr.2: 156329809-156329809 on Build GRCh38
rs762411697 Chr.2: 156329740-156329740 on Build GRCh38
rs750802450 Chr.2: 156329768-156329768 on Build GRCh38
rs781123697 Chr.2: 156329693-156329693 on Build GRCh38
rs771851467 Chr.2: 156329713-156329713 on Build GRCh38
rs755138263 Chr.2: 156329800-156329800 on Build GRCh38
rs201350514 Chr.2: 156329843-156329843 on Build GRCh38
rs757243782 Chr.2: 156329822-156329822 on Build GRCh38
rs747551776 Chr.2: 156329862-156329862 on Build GRCh38
rs751837915 Chr.2: 156329789-156329789 on Build GRCh38
rs776226389 Chr.2: 156329835-156329835 on Build GRCh38
rs771021450 Chr.2: 156329847-156329847 on Build GRCh38
rs575027195 Chr.2: 156329727-156329727 on Build GRCh38
rs763438209 Chr.2: 156329868-156329868 on Build GRCh38
rs554983764 Chr.2: 156329720-156329720 on Build GRCh38
rs752681839 Chr.2: 156329819-156329819 on Build GRCh38
rs562576602 Chr.2: 156329851-156329851 on Build GRCh38
rs771349174 Chr.2: 156329821-156329823 on Build GRCh38
rs368458999 Chr.2: 156329876-156329876 on Build GRCh38
rs758640224 Chr.2: 156329665-156329665 on Build GRCh38
rs749779855 Chr.2: 156329751-156329752 on Build GRCh38
rs775346380 Chr.2: 156329716-156329716 on Build GRCh38
rs770340351 Chr.2: 156329863-156329863 on Build GRCh38
rs182148225 Chr.2: 156329698-156329698 on Build GRCh38
rs755153632 Chr.2: 156329683-156329683 on Build GRCh38
rs758322608 Chr.2: 156329828-156329828 on Build GRCh38
rs751643950 Chr.2: 156329682-156329682 on Build GRCh38
rs370967236 Chr.2: 156329709-156329709 on Build GRCh38
rs200471574 Chr.2: 156329770-156329770 on Build GRCh38
rs150739032 Chr.2: 156329834-156329834 on Build GRCh38
rs201399912 Chr.2: 156329827-156329827 on Build GRCh38
rs797046133 Chr.2: 156329830-156329830 on Build GRCh38
rs943205037 Chr.2: 156329815-156329815 on Build GRCh38
rs958068998 Chr.2: 156329689-156329689 on Build GRCh38
rs779969826 Chr.2: 156329829-156329829 on Build GRCh38

View Full Product Details