Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57208986-57209259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSPRY1
Entrez Gene ID
89970
Gene Name
ring finger and SPRY domain containing 1
Gene Aliases
SEMDFA
UniGene
Hs.460885
Gene Chromosome Location
Chr.16: 57186284-57240475 on Build GRCh38
Transcripts
NM_001305163.1 NM_001305164.1 NM_133368.2 XM_005256220.1 XM_011523427.1 XM_011523428.1 XM_011523430.1 XM_017023844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609444 c.424G>T p.V142F Chr.16: 57209095-57209095 on Build GRCh38
398948 c.460C>G p.P154A Chr.16: 57209131-57209131 on Build GRCh38
6286627 c.486G>T p.L162F Chr.16: 57209157-57209157 on Build GRCh38
703353 c.444T>G p.V148V Chr.16: 57209115-57209115 on Build GRCh38
1128921 c.482C>T p.T161I Chr.16: 57209153-57209153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775692677 Chr.16: 57209061-57209061 on Build GRCh38
rs770955562 Chr.16: 57209203-57209203 on Build GRCh38
rs889502510 Chr.16: 57209105-57209105 on Build GRCh38
rs371561971 Chr.16: 57209198-57209198 on Build GRCh38
rs764258700 Chr.16: 57209077-57209077 on Build GRCh38
rs757822893 Chr.16: 57209092-57209092 on Build GRCh38
rs767751415 Chr.16: 57209141-57209141 on Build GRCh38
rs146849995 Chr.16: 57209079-57209079 on Build GRCh38
rs140565458 Chr.16: 57209176-57209176 on Build GRCh38
rs112596141 Chr.16: 57209046-57209046 on Build GRCh38
rs747819229 Chr.16: 57209166-57209166 on Build GRCh38
rs750879821 Chr.16: 57209154-57209154 on Build GRCh38
rs375654242 Chr.16: 57209070-57209070 on Build GRCh38
rs111967613 Chr.16: 57209021-57209021 on Build GRCh38
rs757803911 Chr.16: 57209172-57209172 on Build GRCh38
rs781319159 Chr.16: 57209127-57209128 on Build GRCh38
rs1005275105 Chr.16: 57209150-57209150 on Build GRCh38
rs778644181 Chr.16: 57209161-57209161 on Build GRCh38
rs754676235 Chr.16: 57209155-57209155 on Build GRCh38
rs755251786 Chr.16: 57209055-57209057 on Build GRCh38
rs867931414 Chr.16: 57209140-57209140 on Build GRCh38
rs371095165 Chr.16: 57209226-57209226 on Build GRCh38
rs867867549 Chr.16: 57209095-57209095 on Build GRCh38
rs745595531 Chr.16: 57209213-57209213 on Build GRCh38
rs776405694 Chr.16: 57209204-57209204 on Build GRCh38
rs901126487 Chr.16: 57209025-57209025 on Build GRCh38
rs147902966 Chr.16: 57209057-57209057 on Build GRCh38
rs374482375 Chr.16: 57209231-57209231 on Build GRCh38
rs572368101 Chr.16: 57209015-57209015 on Build GRCh38

View Full Product Details