Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 113878093-113878354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP1R3A
Entrez Gene ID
5506
Gene Name
protein phosphatase 1 regulatory subunit 3A
Gene Aliases
GM, PP1G, PPP1R3
UniGene
Hs.458309
Gene Chromosome Location
Chr.7: 113876827-113919094 on Build GRCh38
Transcripts
NM_002711.3 XM_005250473.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3877344 c.2779A>G p.T927A Chr.7: 113878313-113878313 on Build GRCh38
3632032 c.2930C>T p.S977F Chr.7: 113878162-113878162 on Build GRCh38
85068 c.2916A>C p.K972N Chr.7: 113878176-113878176 on Build GRCh38
3366729 c.2760T>C p.H920H Chr.7: 113878332-113878332 on Build GRCh38
3632030 c.2949G>A p.V983V Chr.7: 113878143-113878143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56057003 Chr.7: 113878256-113878256 on Build GRCh38
rs557251409 Chr.7: 113878145-113878145 on Build GRCh38
rs144835173 Chr.7: 113878241-113878241 on Build GRCh38
rs35449651 Chr.7: 113878300-113878300 on Build GRCh38
rs945069861 Chr.7: 113878297-113878297 on Build GRCh38
rs773383668 Chr.7: 113878125-113878125 on Build GRCh38
rs776981584 Chr.7: 113878159-113878159 on Build GRCh38
rs764517275 Chr.7: 113878318-113878318 on Build GRCh38
rs752857671 Chr.7: 113878237-113878237 on Build GRCh38
rs936892986 Chr.7: 113878276-113878276 on Build GRCh38
rs150965727 Chr.7: 113878248-113878248 on Build GRCh38
rs555197474 Chr.7: 113878273-113878273 on Build GRCh38
rs776699901 Chr.7: 113878298-113878298 on Build GRCh38
rs140451975 Chr.7: 113878237-113878237 on Build GRCh38
rs913567226 Chr.7: 113878290-113878290 on Build GRCh38
rs1016207297 Chr.7: 113878202-113878202 on Build GRCh38
rs989753229 Chr.7: 113878281-113878281 on Build GRCh38
rs775784657 Chr.7: 113878303-113878303 on Build GRCh38
rs148408134 Chr.7: 113878207-113878207 on Build GRCh38
rs764609245 Chr.7: 113878211-113878211 on Build GRCh38
rs771104644 Chr.7: 113878289-113878289 on Build GRCh38
rs772082412 Chr.7: 113878271-113878271 on Build GRCh38
rs374950521 Chr.7: 113878120-113878120 on Build GRCh38
rs138771673 Chr.7: 113878270-113878270 on Build GRCh38
rs749876989 Chr.7: 113878231-113878231 on Build GRCh38
rs770034298 Chr.7: 113878302-113878302 on Build GRCh38
rs892186853 Chr.7: 113878194-113878194 on Build GRCh38
rs569568096 Chr.7: 113878163-113878163 on Build GRCh38
rs376809464 Chr.7: 113878269-113878269 on Build GRCh38
rs937836738 Chr.7: 113878319-113878319 on Build GRCh38
rs754703917 Chr.7: 113878249-113878249 on Build GRCh38
rs779568010 Chr.7: 113878243-113878243 on Build GRCh38
rs770995700 Chr.7: 113878148-113878148 on Build GRCh38
rs868268486 Chr.7: 113878328-113878328 on Build GRCh38
rs369050094 Chr.7: 113878169-113878169 on Build GRCh38
rs202100582 Chr.7: 113878229-113878229 on Build GRCh38
rs747057183 Chr.7: 113878287-113878287 on Build GRCh38
rs1006150256 Chr.7: 113878130-113878130 on Build GRCh38
rs765683138 Chr.7: 113878174-113878174 on Build GRCh38
rs373483153 Chr.7: 113878313-113878313 on Build GRCh38
rs538868155 Chr.7: 113878142-113878142 on Build GRCh38
rs753171269 Chr.7: 113878197-113878197 on Build GRCh38

View Full Product Details