Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 63566476-63566749 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINB12
Entrez Gene ID
89777
Gene Name
serpin family B member 12
Gene Aliases
YUKOPIN
UniGene
Hs.348541
Gene Chromosome Location
Chr.18: 63519164-63569329 on Build GRCh38
Transcripts
NM_001307928.1 NM_080474.2 XM_005266778.3 XM_011526246.1 XM_011526248.1 XM_011526249.1 XM_017026067.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4416877 c.881C>T p.S294L Chr.18: 63566674-63566674 on Build GRCh38
438310 c.825A>T p.K275N Chr.18: 63566618-63566618 on Build GRCh38
4769601 c.855C>A p.S285R Chr.18: 63566648-63566648 on Build GRCh38
6557110 c.909C>T p.T303T Chr.18: 63566702-63566702 on Build GRCh38
367197 c.901C>A p.R301R Chr.18: 63566694-63566694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs184882895 Chr.18: 63566623-63566623 on Build GRCh38
rs749545 Chr.18: 63566579-63566579 on Build GRCh38
rs764603573 Chr.18: 63566564-63566564 on Build GRCh38
rs537991372 Chr.18: 63566552-63566552 on Build GRCh38
rs768414616 Chr.18: 63566622-63566622 on Build GRCh38
rs534127193 Chr.18: 63566626-63566626 on Build GRCh38
rs748071911 Chr.18: 63566681-63566681 on Build GRCh38
rs760237124 Chr.18: 63566634-63566634 on Build GRCh38
rs781084929 Chr.18: 63566569-63566569 on Build GRCh38
rs749477032 Chr.18: 63566694-63566694 on Build GRCh38
rs997553682 Chr.18: 63566533-63566533 on Build GRCh38
rs766052951 Chr.18: 63566635-63566635 on Build GRCh38
rs377654949 Chr.18: 63566602-63566602 on Build GRCh38
rs201900217 Chr.18: 63566675-63566675 on Build GRCh38
rs909715911 Chr.18: 63566713-63566713 on Build GRCh38
rs541425993 Chr.18: 63566696-63566696 on Build GRCh38
rs746304051 Chr.18: 63566689-63566690 on Build GRCh38
rs78848883 Chr.18: 63566537-63566537 on Build GRCh38
rs1018230461 Chr.18: 63566502-63566502 on Build GRCh38
rs966803978 Chr.18: 63566555-63566555 on Build GRCh38
rs781600809 Chr.18: 63566671-63566672 on Build GRCh38
rs188221723 Chr.18: 63566628-63566628 on Build GRCh38
rs780254213 Chr.18: 63566591-63566591 on Build GRCh38
rs202021125 Chr.18: 63566594-63566594 on Build GRCh38
rs61751994 Chr.18: 63566674-63566674 on Build GRCh38
rs539712383 Chr.18: 63566503-63566503 on Build GRCh38
rs771835916 Chr.18: 63566708-63566708 on Build GRCh38
rs755221725 Chr.18: 63566695-63566695 on Build GRCh38
rs374065223 Chr.18: 63566706-63566706 on Build GRCh38
rs768076403 Chr.18: 63566677-63566677 on Build GRCh38
rs180922662 Chr.18: 63566683-63566683 on Build GRCh38
rs757816373 Chr.18: 63566568-63566568 on Build GRCh38
rs367650019 Chr.18: 63566709-63566709 on Build GRCh38
rs770534987 Chr.18: 63566716-63566716 on Build GRCh38
rs2077549 Chr.18: 63566506-63566506 on Build GRCh38
rs747000331 Chr.18: 63566715-63566715 on Build GRCh38
rs764515832 Chr.18: 63566673-63566673 on Build GRCh38
rs780152976 Chr.18: 63566687-63566687 on Build GRCh38
rs145034398 Chr.18: 63566625-63566625 on Build GRCh38
rs35352345 Chr.18: 63566659-63566659 on Build GRCh38
rs935125785 Chr.18: 63566704-63566704 on Build GRCh38
rs759303030 Chr.18: 63566660-63566660 on Build GRCh38
rs747036721 Chr.18: 63566581-63566581 on Build GRCh38

View Full Product Details