Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50873387-50873634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK2
Entrez Gene ID
3817
Gene Name
kallikrein related peptidase 2
Gene Aliases
KLK2A2, hGK-1, hK2
UniGene
Hs.515560
Gene Chromosome Location
Chr.19: 50873433-50880567 on Build GRCh38
Transcripts
NM_001002231.2 NM_001256080.1 NM_005551.4 NR_045762.1 NR_045763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4285830 c.20C>T p.S7F Chr.19: 50873493-50873493 on Build GRCh38
5417430 c.31T>C p.S11P Chr.19: 50873504-50873504 on Build GRCh38
6860369 c.37G>C p.G13R Chr.19: 50873510-50873510 on Build GRCh38
7374233 c.6G>T p.W2C Chr.19: 50873479-50873479 on Build GRCh38
7374234 c.6G>T p.W2C Chr.19: 50873479-50873479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377048033 Chr.19: 50873526-50873526 on Build GRCh38
rs749499828 Chr.19: 50873500-50873500 on Build GRCh38
rs373131194 Chr.19: 50873449-50873449 on Build GRCh38
rs371299015 Chr.19: 50873550-50873550 on Build GRCh38
rs925327214 Chr.19: 50873607-50873607 on Build GRCh38
rs891363873 Chr.19: 50873483-50873483 on Build GRCh38
rs760140166 Chr.19: 50873537-50873537 on Build GRCh38
rs997395630 Chr.19: 50873429-50873429 on Build GRCh38
rs894073945 Chr.19: 50873474-50873474 on Build GRCh38
rs62113073 Chr.19: 50873581-50873581 on Build GRCh38
rs111990959 Chr.19: 50873601-50873601 on Build GRCh38
rs377554986 Chr.19: 50873450-50873450 on Build GRCh38
rs367898018 Chr.19: 50873466-50873466 on Build GRCh38
rs755348099 Chr.19: 50873557-50873557 on Build GRCh38
rs372599210 Chr.19: 50873494-50873494 on Build GRCh38
rs1043801207 Chr.19: 50873416-50873416 on Build GRCh38
rs371149173 Chr.19: 50873455-50873455 on Build GRCh38
rs781691944 Chr.19: 50873488-50873488 on Build GRCh38
rs764583501 Chr.19: 50873468-50873468 on Build GRCh38
rs774136702 Chr.19: 50873505-50873505 on Build GRCh38
rs753506515 Chr.19: 50873440-50873440 on Build GRCh38
rs187716365 Chr.19: 50873491-50873491 on Build GRCh38
rs34652810 Chr.19: 50873443-50873444 on Build GRCh38
rs756208467 Chr.19: 50873571-50873571 on Build GRCh38
rs112897428 Chr.19: 50873497-50873497 on Build GRCh38
rs766118997 Chr.19: 50873549-50873549 on Build GRCh38
rs780581667 Chr.19: 50873456-50873456 on Build GRCh38
rs368057223 Chr.19: 50873548-50873548 on Build GRCh38
rs369907394 Chr.19: 50873430-50873430 on Build GRCh38
rs777336236 Chr.19: 50873434-50873434 on Build GRCh38
rs754914185 Chr.19: 50873588-50873588 on Build GRCh38
rs755539982 Chr.19: 50873427-50873427 on Build GRCh38
rs146007033 Chr.19: 50873503-50873503 on Build GRCh38
rs759320056 Chr.19: 50873615-50873615 on Build GRCh38
rs759286391 Chr.19: 50873524-50873524 on Build GRCh38
rs368076226 Chr.19: 50873462-50873462 on Build GRCh38
rs775472259 Chr.19: 50873529-50873529 on Build GRCh38
rs751998602 Chr.19: 50873552-50873552 on Build GRCh38
rs201923328 Chr.19: 50873566-50873566 on Build GRCh38
rs763653675 Chr.19: 50873546-50873546 on Build GRCh38
rs781590914 Chr.19: 50873564-50873564 on Build GRCh38
rs979527806 Chr.19: 50873604-50873604 on Build GRCh38
rs779473504 Chr.19: 50873549-50873549 on Build GRCh38

View Full Product Details