Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68361668-68361895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMPD3
Entrez Gene ID
55512
Gene Name
sphingomyelin phosphodiesterase 3
Gene Aliases
NSMASE2
UniGene
Hs.368421
Gene Chromosome Location
Chr.16: 68358327-68448703 on Build GRCh38
Transcripts
NM_018667.3 XM_005256031.3 XM_005256032.3 XM_011523207.1 XM_011523208.2 XM_011523209.2 XM_017023405.1 XM_017023406.1 XM_017023407.1 XM_017023408.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1379108 c.1729G>A p.G577S Chr.16: 68361740-68361740 on Build GRCh38
6785639 c.1769C>T p.S590L Chr.16: 68361700-68361700 on Build GRCh38
3957748 c.1793T>C p.L598P Chr.16: 68361676-68361676 on Build GRCh38
6785633 c.1733G>A p.R578H Chr.16: 68361736-68361736 on Build GRCh38
1226898 c.1781G>T p.G594V Chr.16: 68361688-68361688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531491814 Chr.16: 68361773-68361773 on Build GRCh38
rs777091338 Chr.16: 68361798-68361798 on Build GRCh38
rs752784495 Chr.16: 68361741-68361741 on Build GRCh38
rs1016674358 Chr.16: 68361720-68361720 on Build GRCh38
rs750352174 Chr.16: 68361751-68361751 on Build GRCh38
rs150682635 Chr.16: 68361753-68361753 on Build GRCh38
rs779744838 Chr.16: 68361705-68361705 on Build GRCh38
rs766847055 Chr.16: 68361726-68361726 on Build GRCh38
rs78777441 Chr.16: 68361698-68361698 on Build GRCh38
rs907218153 Chr.16: 68361819-68361819 on Build GRCh38
rs779797953 Chr.16: 68361756-68361756 on Build GRCh38
rs775558692 Chr.16: 68361811-68361811 on Build GRCh38
rs771472010 Chr.16: 68361771-68361771 on Build GRCh38
rs754619056 Chr.16: 68361766-68361766 on Build GRCh38
rs936181761 Chr.16: 68361764-68361764 on Build GRCh38
rs377365842 Chr.16: 68361721-68361721 on Build GRCh38
rs761572378 Chr.16: 68361804-68361804 on Build GRCh38
rs56899435 Chr.16: 68361772-68361772 on Build GRCh38
rs879004023 Chr.16: 68361693-68361693 on Build GRCh38
rs956723917 Chr.16: 68361826-68361826 on Build GRCh38
rs776627090 Chr.16: 68361764-68361765 on Build GRCh38
rs772561670 Chr.16: 68361723-68361723 on Build GRCh38
rs764477262 Chr.16: 68361859-68361859 on Build GRCh38
rs765258721 Chr.16: 68361740-68361740 on Build GRCh38
rs759871907 Chr.16: 68361799-68361799 on Build GRCh38
rs767546455 Chr.16: 68361749-68361749 on Build GRCh38
rs763390470 Chr.16: 68361841-68361841 on Build GRCh38
rs757148334 Chr.16: 68361747-68361747 on Build GRCh38
rs975045171 Chr.16: 68361867-68361867 on Build GRCh38
rs201079929 Chr.16: 68361715-68361715 on Build GRCh38
rs990223835 Chr.16: 68361770-68361770 on Build GRCh38
rs955621644 Chr.16: 68361744-68361744 on Build GRCh38
rs773056401 Chr.16: 68361686-68361686 on Build GRCh38
rs773861235 Chr.16: 68361716-68361716 on Build GRCh38
rs759814899 Chr.16: 68361737-68361737 on Build GRCh38
rs866248972 Chr.16: 68361843-68361843 on Build GRCh38
rs550095026 Chr.16: 68361806-68361806 on Build GRCh38
rs529093102 Chr.16: 68361858-68361858 on Build GRCh38
rs1045705981 Chr.16: 68361797-68361797 on Build GRCh38
rs533134055 Chr.16: 68361825-68361825 on Build GRCh38
rs747634496 Chr.16: 68361768-68361768 on Build GRCh38
rs568114105 Chr.16: 68361835-68361835 on Build GRCh38
rs368768529 Chr.16: 68361779-68361779 on Build GRCh38
rs540263654 Chr.16: 68361849-68361849 on Build GRCh38
rs1038168521 Chr.16: 68361857-68361857 on Build GRCh38
rs971392882 Chr.16: 68361792-68361792 on Build GRCh38
rs745601859 Chr.16: 68361700-68361700 on Build GRCh38
rs780882838 Chr.16: 68361699-68361699 on Build GRCh38
rs773464159 Chr.16: 68361725-68361725 on Build GRCh38
rs370760321 Chr.16: 68361758-68361758 on Build GRCh38
rs778289387 Chr.16: 68361767-68361767 on Build GRCh38
rs777034210 Chr.16: 68361736-68361736 on Build GRCh38
rs560204511 Chr.16: 68361702-68361702 on Build GRCh38
rs748904266 Chr.16: 68361708-68361708 on Build GRCh38
rs374101126 Chr.16: 68361791-68361791 on Build GRCh38

View Full Product Details