Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40530140-40530399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF621
Entrez Gene ID
285268
Gene Name
zinc finger protein 621
Gene Aliases
-
UniGene
Hs.19977
Gene Chromosome Location
Chr.3: 40523854-40539794 on Build GRCh38
Transcripts
NM_001098414.2 NM_001287245.1 NM_198484.4 XM_005265079.3 XM_005265080.3 XM_006713114.3 XM_011533610.2 XM_017006226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6831827 c.155C>T p.A52V Chr.3: 40530212-40530212 on Build GRCh38
3593963 c.189C>T p.H63H Chr.3: 40530246-40530246 on Build GRCh38
3593964 c.224C>T p.P75L Chr.3: 40530281-40530281 on Build GRCh38
730876 c.195G>A p.E65E Chr.3: 40530252-40530252 on Build GRCh38
730877 c.180G>A p.L60L Chr.3: 40530237-40530237 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767747596 Chr.3: 40530188-40530188 on Build GRCh38
rs757511659 Chr.3: 40530333-40530333 on Build GRCh38
rs991797446 Chr.3: 40530334-40530334 on Build GRCh38
rs770964757 Chr.3: 40530292-40530292 on Build GRCh38
rs200611775 Chr.3: 40530316-40530316 on Build GRCh38
rs982991987 Chr.3: 40530222-40530222 on Build GRCh38
rs370665011 Chr.3: 40530176-40530176 on Build GRCh38
rs745954541 Chr.3: 40530306-40530306 on Build GRCh38
rs754762728 Chr.3: 40530368-40530368 on Build GRCh38
rs200153819 Chr.3: 40530362-40530362 on Build GRCh38
rs775872579 Chr.3: 40530212-40530212 on Build GRCh38
rs771054813 Chr.3: 40530172-40530172 on Build GRCh38
rs747136554 Chr.3: 40530291-40530291 on Build GRCh38
rs758156140 Chr.3: 40530182-40530182 on Build GRCh38
rs367913818 Chr.3: 40530304-40530304 on Build GRCh38
rs779596362 Chr.3: 40530267-40530267 on Build GRCh38
rs141664376 Chr.3: 40530170-40530170 on Build GRCh38
rs374336887 Chr.3: 40530242-40530242 on Build GRCh38
rs764614136 Chr.3: 40530327-40530327 on Build GRCh38
rs750014697 Chr.3: 40530262-40530262 on Build GRCh38
rs762402175 Chr.3: 40530360-40530360 on Build GRCh38
rs749901205 Chr.3: 40530351-40530351 on Build GRCh38
rs764910226 Chr.3: 40530255-40530255 on Build GRCh38
rs761046676 Chr.3: 40530320-40530320 on Build GRCh38
rs751227345 Chr.3: 40530367-40530367 on Build GRCh38
rs536305510 Chr.3: 40530315-40530315 on Build GRCh38
rs199764985 Chr.3: 40530279-40530279 on Build GRCh38
rs756816845 Chr.3: 40530250-40530250 on Build GRCh38
rs779699569 Chr.3: 40530272-40530273 on Build GRCh38
rs558943324 Chr.3: 40530364-40530364 on Build GRCh38
rs758120993 Chr.3: 40530265-40530265 on Build GRCh38
rs746779831 Chr.3: 40530269-40530269 on Build GRCh38
rs972175292 Chr.3: 40530223-40530223 on Build GRCh38
rs917618449 Chr.3: 40530363-40530363 on Build GRCh38
rs764634165 Chr.3: 40530225-40530225 on Build GRCh38
rs377314913 Chr.3: 40530323-40530323 on Build GRCh38
rs1016496365 Chr.3: 40530192-40530192 on Build GRCh38
rs370605918 Chr.3: 40530213-40530213 on Build GRCh38
rs573160555 Chr.3: 40530328-40530328 on Build GRCh38
rs762452043 Chr.3: 40530331-40530331 on Build GRCh38
rs149905428 Chr.3: 40530270-40530270 on Build GRCh38
rs368663755 Chr.3: 40530171-40530171 on Build GRCh38

View Full Product Details