Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196665468-196665739 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC150
Entrez Gene ID
284992
Gene Name
coiled-coil domain containing 150
Gene Aliases
-
UniGene
Hs.132519
Gene Chromosome Location
Chr.2: 196639632-196733668 on Build GRCh38
Transcripts
NM_001080539.1 XM_006712438.3 XM_011510985.2 XM_011510988.2 XM_017003861.1 XM_017003862.1 XM_017003863.1 XM_017003864.1 XM_017003865.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7225890 c.722T>C p.I241T Chr.2: 196665643-196665643 on Build GRCh38
3575971 c.686C>T p.S229F Chr.2: 196665607-196665607 on Build GRCh38
1404232 c.657A>G p.Q219Q Chr.2: 196665578-196665578 on Build GRCh38
1014405 c.754C>T p.R252* Chr.2: 196665675-196665675 on Build GRCh38
7416036 c.747G>T p.E249D Chr.2: 196665668-196665668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141465191 Chr.2: 196665503-196665503 on Build GRCh38
rs749977575 Chr.2: 196665651-196665651 on Build GRCh38
rs759850028 Chr.2: 196665628-196665628 on Build GRCh38
rs1046187444 Chr.2: 196665629-196665629 on Build GRCh38
rs760153085 Chr.2: 196665662-196665662 on Build GRCh38
rs536815408 Chr.2: 196665680-196665680 on Build GRCh38
rs763513580 Chr.2: 196665642-196665642 on Build GRCh38
rs199618545 Chr.2: 196665618-196665618 on Build GRCh38
rs776973958 Chr.2: 196665634-196665634 on Build GRCh38
rs74454431 Chr.2: 196665578-196665578 on Build GRCh38
rs372569306 Chr.2: 196665553-196665553 on Build GRCh38
rs748028276 Chr.2: 196665594-196665594 on Build GRCh38
rs548240746 Chr.2: 196665676-196665676 on Build GRCh38
rs772290944 Chr.2: 196665599-196665599 on Build GRCh38
rs765966998 Chr.2: 196665663-196665663 on Build GRCh38
rs145159886 Chr.2: 196665524-196665524 on Build GRCh38
rs778057117 Chr.2: 196665603-196665603 on Build GRCh38
rs759924592 Chr.2: 196665522-196665522 on Build GRCh38
rs190297893 Chr.2: 196665641-196665641 on Build GRCh38
rs202231571 Chr.2: 196665643-196665643 on Build GRCh38
rs764279963 Chr.2: 196665600-196665601 on Build GRCh38
rs756764544 Chr.2: 196665539-196665539 on Build GRCh38
rs1023385019 Chr.2: 196665610-196665610 on Build GRCh38
rs747079980 Chr.2: 196665607-196665607 on Build GRCh38
rs1025284090 Chr.2: 196665533-196665533 on Build GRCh38
rs749926024 Chr.2: 196665547-196665547 on Build GRCh38
rs776865395 Chr.2: 196665626-196665626 on Build GRCh38
rs374830934 Chr.2: 196665552-196665558 on Build GRCh38
rs749083634 Chr.2: 196665559-196665559 on Build GRCh38
rs373966441 Chr.2: 196665683-196665683 on Build GRCh38
rs568815221 Chr.2: 196665592-196665592 on Build GRCh38
rs144653156 Chr.2: 196665671-196665671 on Build GRCh38
rs766960775 Chr.2: 196665542-196665542 on Build GRCh38
rs762203687 Chr.2: 196665689-196665691 on Build GRCh38
rs752136982 Chr.2: 196665513-196665513 on Build GRCh38
rs898040336 Chr.2: 196665514-196665514 on Build GRCh38
rs930238096 Chr.2: 196665627-196665627 on Build GRCh38
rs80092902 Chr.2: 196665690-196665690 on Build GRCh38
rs1034946203 Chr.2: 196665655-196665655 on Build GRCh38
rs770129722 Chr.2: 196665637-196665637 on Build GRCh38
rs368240597 Chr.2: 196665540-196665540 on Build GRCh38
rs375529700 Chr.2: 196665526-196665526 on Build GRCh38
rs1005327986 Chr.2: 196665595-196665595 on Build GRCh38
rs568066863 Chr.2: 196665678-196665678 on Build GRCh38
rs1038531122 Chr.2: 196665623-196665623 on Build GRCh38

View Full Product Details