Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62839403-62839674 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR74
Entrez Gene ID
54663
Gene Name
WD repeat domain 74
Gene Aliases
-
UniGene
Hs.654620
Gene Chromosome Location
Chr.11: 62832905-62841809 on Build GRCh38
Transcripts
NM_001307977.1 NM_018093.3 XM_005274055.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322287 c.21C>G p.R7R Chr.11: 62839550-62839550 on Build GRCh38
5623421 c.64+2T>G p.? Chr.11: 62839505-62839505 on Build GRCh38
3986310 c.60G>C p.L20F Chr.11: 62839511-62839511 on Build GRCh38
2039859 c.54G>T p.G18G Chr.11: 62839517-62839517 on Build GRCh38
6265511 c.79C>A p.R27R Chr.11: 62839414-62839414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751927322 Chr.11: 62839514-62839514 on Build GRCh38
rs908493827 Chr.11: 62839562-62839562 on Build GRCh38
rs765480892 Chr.11: 62839554-62839556 on Build GRCh38
rs747332581 Chr.11: 62839430-62839430 on Build GRCh38
rs748874864 Chr.11: 62839442-62839442 on Build GRCh38
rs1049492574 Chr.11: 62839649-62839649 on Build GRCh38
rs761398726 Chr.11: 62839462-62839462 on Build GRCh38
rs778537136 Chr.11: 62839558-62839558 on Build GRCh38
rs781774997 Chr.11: 62839524-62839524 on Build GRCh38
rs765155158 Chr.11: 62839552-62839552 on Build GRCh38
rs776515689 Chr.11: 62839487-62839487 on Build GRCh38
rs771719739 Chr.11: 62839467-62839467 on Build GRCh38
rs150613372 Chr.11: 62839492-62839492 on Build GRCh38
rs775446241 Chr.11: 62839560-62839560 on Build GRCh38
rs774526347 Chr.11: 62839602-62839602 on Build GRCh38
rs772893432 Chr.11: 62839479-62839479 on Build GRCh38
rs965722772 Chr.11: 62839453-62839453 on Build GRCh38
rs1055033034 Chr.11: 62839464-62839464 on Build GRCh38
rs771676502 Chr.11: 62839542-62839542 on Build GRCh38
rs770688561 Chr.11: 62839549-62839549 on Build GRCh38
rs7102434 Chr.11: 62839596-62839596 on Build GRCh38
rs754333816 Chr.11: 62839517-62839517 on Build GRCh38
rs752782215 Chr.11: 62839500-62839500 on Build GRCh38
rs768761160 Chr.11: 62839599-62839599 on Build GRCh38
rs547512268 Chr.11: 62839644-62839644 on Build GRCh38
rs200010672 Chr.11: 62839563-62839563 on Build GRCh38
rs371820946 Chr.11: 62839484-62839484 on Build GRCh38
rs770529340 Chr.11: 62839590-62839590 on Build GRCh38
rs764291852 Chr.11: 62839564-62839564 on Build GRCh38
rs114810174 Chr.11: 62839550-62839550 on Build GRCh38
rs776414054 Chr.11: 62839591-62839591 on Build GRCh38
rs764320206 Chr.11: 62839506-62839506 on Build GRCh38
rs747632170 Chr.11: 62839541-62839541 on Build GRCh38
rs777336558 Chr.11: 62839588-62839588 on Build GRCh38
rs746844061 Chr.11: 62839548-62839548 on Build GRCh38
rs369606671 Chr.11: 62839592-62839592 on Build GRCh38
rs768253603 Chr.11: 62839459-62839459 on Build GRCh38
rs759766180 Chr.11: 62839426-62839426 on Build GRCh38
rs774105076 Chr.11: 62839460-62839460 on Build GRCh38
rs368356605 Chr.11: 62839445-62839445 on Build GRCh38
rs750781500 Chr.11: 62839577-62839577 on Build GRCh38
rs368591562 Chr.11: 62839539-62839539 on Build GRCh38
rs771714689 Chr.11: 62839618-62839618 on Build GRCh38
rs769677990 Chr.11: 62839593-62839593 on Build GRCh38
rs768018914 Chr.11: 62839569-62839569 on Build GRCh38
rs781761716 Chr.11: 62839441-62839441 on Build GRCh38
rs200644559 Chr.11: 62839480-62839480 on Build GRCh38
rs545160528 Chr.11: 62839566-62839566 on Build GRCh38
rs772909911 Chr.11: 62839547-62839547 on Build GRCh38
rs746358516 Chr.11: 62839526-62839526 on Build GRCh38
rs762161451 Chr.11: 62839619-62839619 on Build GRCh38
rs757694760 Chr.11: 62839516-62839516 on Build GRCh38
rs746684567 Chr.11: 62839589-62839589 on Build GRCh38
rs74789950 Chr.11: 62839435-62839435 on Build GRCh38
rs758552593 Chr.11: 62839505-62839505 on Build GRCh38
rs747674525 Chr.11: 62839586-62839586 on Build GRCh38
rs756697957 Chr.11: 62839580-62839580 on Build GRCh38
rs527331938 Chr.11: 62839597-62839597 on Build GRCh38
rs765321626 Chr.11: 62839498-62839498 on Build GRCh38
rs374910485 Chr.11: 62839584-62839584 on Build GRCh38

View Full Product Details