Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19852123-19852356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZMYM5
Entrez Gene ID
9205
Gene Name
zinc finger MYM-type containing 5
Gene Aliases
HSPC050, MYM, ZNF198L1, ZNF237
UniGene
Hs.530988
Gene Chromosome Location
Chr.13: 19823482-19863636 on Build GRCh38
Transcripts
NM_001039649.2 NM_001039650.2 NM_001142684.1 XM_005266593.3 XM_005266594.2 XM_006719894.2 XM_011535309.1 XM_011535310.1 XM_017020849.1 XM_017020850.1 XM_017020851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6827575 c.16G>A p.V6M Chr.13: 19852165-19852165 on Build GRCh38
5842249 c.46C>T p.P16S Chr.13: 19852135-19852135 on Build GRCh38
6827574 c.16G>A p.V6M Chr.13: 19852165-19852165 on Build GRCh38
6553828 c.49G>A p.A17T Chr.13: 19852132-19852132 on Build GRCh38
6553827 c.49G>A p.A17T Chr.13: 19852132-19852132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754205783 Chr.13: 19852228-19852228 on Build GRCh38
rs779630978 Chr.13: 19852153-19852154 on Build GRCh38
rs548909144 Chr.13: 19852268-19852268 on Build GRCh38
rs760102145 Chr.13: 19852232-19852232 on Build GRCh38
rs904990333 Chr.13: 19852197-19852197 on Build GRCh38
rs779791460 Chr.13: 19852151-19852151 on Build GRCh38
rs749266505 Chr.13: 19852208-19852211 on Build GRCh38
rs766879689 Chr.13: 19852227-19852227 on Build GRCh38
rs753224706 Chr.13: 19852172-19852172 on Build GRCh38
rs768043082 Chr.13: 19852157-19852157 on Build GRCh38
rs1040164308 Chr.13: 19852185-19852185 on Build GRCh38
rs768424298 Chr.13: 19852157-19852157 on Build GRCh38
rs56100501 Chr.13: 19852241-19852241 on Build GRCh38
rs963948313 Chr.13: 19852208-19852212 on Build GRCh38
rs1002320159 Chr.13: 19852203-19852203 on Build GRCh38
rs1027705312 Chr.13: 19852248-19852248 on Build GRCh38
rs951963166 Chr.13: 19852261-19852261 on Build GRCh38
rs150696327 Chr.13: 19852249-19852249 on Build GRCh38
rs756537381 Chr.13: 19852207-19852208 on Build GRCh38
rs1002083689 Chr.13: 19852282-19852282 on Build GRCh38
rs1008608631 Chr.13: 19852183-19852183 on Build GRCh38
rs141598153 Chr.13: 19852173-19852173 on Build GRCh38
rs897552195 Chr.13: 19852198-19852198 on Build GRCh38
rs370422647 Chr.13: 19852153-19852153 on Build GRCh38
rs761108571 Chr.13: 19852270-19852270 on Build GRCh38
rs1046136797 Chr.13: 19852195-19852196 on Build GRCh38
rs987506139 Chr.13: 19852258-19852258 on Build GRCh38
rs778365384 Chr.13: 19852208-19852208 on Build GRCh38
rs372273523 Chr.13: 19852192-19852192 on Build GRCh38
rs760374694 Chr.13: 19852219-19852219 on Build GRCh38
rs771695981 Chr.13: 19852186-19852186 on Build GRCh38
rs757360290 Chr.13: 19852222-19852222 on Build GRCh38

View Full Product Details