Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44166691-44166940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN11
Entrez Gene ID
11131
Gene Name
calpain 11
Gene Aliases
calpain11
UniGene
Hs.225953
Gene Chromosome Location
Chr.6: 44158811-44184402 on Build GRCh38
Transcripts
NM_007058.3 XM_006714984.1 XM_006714985.1 XM_006714987.1 XM_011514274.1 XM_011514275.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5810792 c.71C>T p.P24L Chr.6: 44166812-44166812 on Build GRCh38
7319946 c.29C>T p.P10L Chr.6: 44166770-44166770 on Build GRCh38
5756880 c.86C>T p.A29V Chr.6: 44166827-44166827 on Build GRCh38
4006525 c.21G>C p.P7P Chr.6: 44166762-44166762 on Build GRCh38
7359417 c.62A>C p.E21A Chr.6: 44166803-44166803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs951199005 Chr.6: 44166918-44166918 on Build GRCh38
rs901085653 Chr.6: 44166827-44166827 on Build GRCh38
rs1011024632 Chr.6: 44166788-44166788 on Build GRCh38
rs755419684 Chr.6: 44166876-44166878 on Build GRCh38
rs530355526 Chr.6: 44166866-44166866 on Build GRCh38
rs188033949 Chr.6: 44166896-44166896 on Build GRCh38
rs540486701 Chr.6: 44166762-44166762 on Build GRCh38
rs901108781 Chr.6: 44166751-44166751 on Build GRCh38
rs1005282966 Chr.6: 44166808-44166808 on Build GRCh38
rs3832440 Chr.6: 44166881-44166883 on Build GRCh38
rs768931952 Chr.6: 44166843-44166861 on Build GRCh38
rs867440318 Chr.6: 44166797-44166797 on Build GRCh38
rs1055229248 Chr.6: 44166768-44166768 on Build GRCh38
rs368698407 Chr.6: 44166838-44166838 on Build GRCh38
rs755679773 Chr.6: 44166769-44166769 on Build GRCh38
rs775423577 Chr.6: 44166850-44166850 on Build GRCh38
rs914503 Chr.6: 44166844-44166844 on Build GRCh38
rs746025988 Chr.6: 44166846-44166846 on Build GRCh38
rs989259470 Chr.6: 44166845-44166845 on Build GRCh38
rs771238539 Chr.6: 44166841-44166841 on Build GRCh38
rs560686072 Chr.6: 44166899-44166899 on Build GRCh38
rs778240948 Chr.6: 44166836-44166836 on Build GRCh38
rs9462967 Chr.6: 44166803-44166803 on Build GRCh38
rs930746410 Chr.6: 44166771-44166771 on Build GRCh38
rs544766080 Chr.6: 44166814-44166814 on Build GRCh38
rs184018090 Chr.6: 44166785-44166785 on Build GRCh38
rs35518505 Chr.6: 44166743-44166744 on Build GRCh38
rs878867230 Chr.6: 44166810-44166810 on Build GRCh38
rs34901210 Chr.6: 44166874-44166876 on Build GRCh38
rs998076587 Chr.6: 44166828-44166828 on Build GRCh38
rs966442986 Chr.6: 44166887-44166887 on Build GRCh38
rs371144537 Chr.6: 44166761-44166761 on Build GRCh38
rs1022047275 Chr.6: 44166871-44166871 on Build GRCh38
rs769933683 Chr.6: 44166849-44166849 on Build GRCh38

View Full Product Details