Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 47501633-47501884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELP6
Entrez Gene ID
54859
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C3orf75, TMEM103
UniGene
Hs.311100
Gene Chromosome Location
Chr.3: 47495640-47513711 on Build GRCh38
Transcripts
NM_001031703.2 XM_005265241.3 XM_011533851.2 XM_011533852.2 XM_011533853.2 XM_011533854.2 XM_011533856.2 XM_011533857.2 XM_017006669.1 XM_017006670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6006934 c.439A>T p.S147C Chr.3: 47501736-47501736 on Build GRCh38
3781823 c.425T>C p.L142S Chr.3: 47501750-47501750 on Build GRCh38
7203840 c.343T>A p.L115M Chr.3: 47501832-47501832 on Build GRCh38
5426894 c.411G>A p.T137T Chr.3: 47501764-47501764 on Build GRCh38
1045629 c.467C>T p.A156V Chr.3: 47501708-47501708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769811559 Chr.3: 47501778-47501778 on Build GRCh38
rs752000466 Chr.3: 47501741-47501741 on Build GRCh38
rs931047050 Chr.3: 47501708-47501708 on Build GRCh38
rs761856562 Chr.3: 47501713-47501713 on Build GRCh38
rs749222312 Chr.3: 47501679-47501679 on Build GRCh38
rs773631436 Chr.3: 47501724-47501724 on Build GRCh38
rs745668948 Chr.3: 47501772-47501772 on Build GRCh38
rs1035573511 Chr.3: 47501743-47501743 on Build GRCh38
rs887545835 Chr.3: 47501811-47501811 on Build GRCh38
rs781335500 Chr.3: 47501775-47501775 on Build GRCh38
rs757210202 Chr.3: 47501771-47501771 on Build GRCh38
rs374797454 Chr.3: 47501839-47501839 on Build GRCh38
rs376912609 Chr.3: 47501707-47501707 on Build GRCh38
rs760033103 Chr.3: 47501808-47501808 on Build GRCh38
rs1006305584 Chr.3: 47501836-47501836 on Build GRCh38
rs763084928 Chr.3: 47501801-47501801 on Build GRCh38
rs774937315 Chr.3: 47501807-47501807 on Build GRCh38
rs773362295 Chr.3: 47501784-47501784 on Build GRCh38
rs148244740 Chr.3: 47501668-47501668 on Build GRCh38
rs777308848 Chr.3: 47501764-47501764 on Build GRCh38
rs771566907 Chr.3: 47501803-47501803 on Build GRCh38
rs753825070 Chr.3: 47501765-47501765 on Build GRCh38
rs956679269 Chr.3: 47501859-47501859 on Build GRCh38
rs531080594 Chr.3: 47501773-47501773 on Build GRCh38
rs993246926 Chr.3: 47501761-47501761 on Build GRCh38
rs367968152 Chr.3: 47501737-47501737 on Build GRCh38
rs778658835 Chr.3: 47501700-47501700 on Build GRCh38
rs748370247 Chr.3: 47501777-47501777 on Build GRCh38
rs181991988 Chr.3: 47501758-47501758 on Build GRCh38
rs763530388 Chr.3: 47501728-47501728 on Build GRCh38
rs770756029 Chr.3: 47501681-47501681 on Build GRCh38
rs748506406 Chr.3: 47501760-47501760 on Build GRCh38
rs755649678 Chr.3: 47501759-47501759 on Build GRCh38
rs542051402 Chr.3: 47501742-47501742 on Build GRCh38
rs767085247 Chr.3: 47501749-47501749 on Build GRCh38
rs769912824 Chr.3: 47501717-47501717 on Build GRCh38
rs201705102 Chr.3: 47501672-47501672 on Build GRCh38
rs745614182 Chr.3: 47501706-47501706 on Build GRCh38
rs368378749 Chr.3: 47501667-47501667 on Build GRCh38
rs776846659 Chr.3: 47501710-47501710 on Build GRCh38

View Full Product Details