Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67281458-67281713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK2
Entrez Gene ID
156
Gene Name
G protein-coupled receptor kinase 2
Gene Aliases
ADRBK1, BARK1, BETA-ARK1
UniGene
Hs.83636
Gene Chromosome Location
Chr.11: 67266420-67286558 on Build GRCh38
Transcripts
NM_001619.3 XM_011544773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1356366 c.662G>A p.C221Y Chr.11: 67281473-67281473 on Build GRCh38
7166944 c.723C>G p.I241M Chr.11: 67281534-67281534 on Build GRCh38
5839361 c.788C>T p.T263M Chr.11: 67281690-67281690 on Build GRCh38
1235741 c.748G>A p.D250N Chr.11: 67281650-67281650 on Build GRCh38
6045638 c.781T>C p.F261L Chr.11: 67281683-67281683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752863003 Chr.11: 67281581-67281581 on Build GRCh38
rs147086415 Chr.11: 67281670-67281670 on Build GRCh38
rs749944374 Chr.11: 67281640-67281641 on Build GRCh38
rs368479232 Chr.11: 67281554-67281554 on Build GRCh38
rs755938304 Chr.11: 67281602-67281602 on Build GRCh38
rs987418340 Chr.11: 67281628-67281628 on Build GRCh38
rs543467610 Chr.11: 67281690-67281690 on Build GRCh38
rs374629054 Chr.11: 67281480-67281480 on Build GRCh38
rs768156866 Chr.11: 67281532-67281532 on Build GRCh38
rs771294109 Chr.11: 67281629-67281629 on Build GRCh38
rs1035537279 Chr.11: 67281504-67281504 on Build GRCh38
rs377245474 Chr.11: 67281622-67281622 on Build GRCh38
rs766995109 Chr.11: 67281625-67281634 on Build GRCh38
rs774022687 Chr.11: 67281597-67281598 on Build GRCh38
rs374203459 Chr.11: 67281639-67281639 on Build GRCh38
rs766767718 Chr.11: 67281568-67281568 on Build GRCh38
rs148898193 Chr.11: 67281679-67281679 on Build GRCh38
rs775843406 Chr.11: 67281641-67281641 on Build GRCh38
rs930238100 Chr.11: 67281625-67281625 on Build GRCh38
rs759787899 Chr.11: 67281635-67281635 on Build GRCh38
rs777192967 Chr.11: 67281627-67281627 on Build GRCh38
rs765352903 Chr.11: 67281577-67281577 on Build GRCh38
rs772631517 Chr.11: 67281546-67281546 on Build GRCh38
rs377123753 Chr.11: 67281682-67281682 on Build GRCh38
rs369783949 Chr.11: 67281623-67281623 on Build GRCh38
rs764274714 Chr.11: 67281646-67281646 on Build GRCh38
rs145638181 Chr.11: 67281535-67281535 on Build GRCh38
rs1023647274 Chr.11: 67281582-67281582 on Build GRCh38
rs374189744 Chr.11: 67281583-67281583 on Build GRCh38
rs760688917 Chr.11: 67281677-67281677 on Build GRCh38
rs748990821 Chr.11: 67281607-67281607 on Build GRCh38
rs370768597 Chr.11: 67281575-67281575 on Build GRCh38
rs576089239 Chr.11: 67281688-67281688 on Build GRCh38
rs776820579 Chr.11: 67281574-67281574 on Build GRCh38
rs923400740 Chr.11: 67281564-67281564 on Build GRCh38
rs761423603 Chr.11: 67281601-67281602 on Build GRCh38
rs748979223 Chr.11: 67281525-67281525 on Build GRCh38
rs61763964 Chr.11: 67281621-67281621 on Build GRCh38
rs770121759 Chr.11: 67281638-67281638 on Build GRCh38
rs751653503 Chr.11: 67281651-67281651 on Build GRCh38
rs777286637 Chr.11: 67281686-67281686 on Build GRCh38
rs745725870 Chr.11: 67281494-67281494 on Build GRCh38
rs753710294 Chr.11: 67281680-67281680 on Build GRCh38
rs758003046 Chr.11: 67281685-67281685 on Build GRCh38
rs767621947 Chr.11: 67281586-67281586 on Build GRCh38
rs763209958 Chr.11: 67281642-67281642 on Build GRCh38
rs781246400 Chr.11: 67281487-67281487 on Build GRCh38
rs777100371 Chr.11: 67281630-67281630 on Build GRCh38
rs200052868 Chr.11: 67281543-67281543 on Build GRCh38
rs201975411 Chr.11: 67281601-67281601 on Build GRCh38
rs779656272 Chr.11: 67281519-67281519 on Build GRCh38
rs770458416 Chr.11: 67281547-67281547 on Build GRCh38
rs372209829 Chr.11: 67281605-67281605 on Build GRCh38
rs947288361 Chr.11: 67281606-67281606 on Build GRCh38
rs771507227 Chr.11: 67281691-67281691 on Build GRCh38
rs773936786 Chr.11: 67281536-67281536 on Build GRCh38
rs755961102 Chr.11: 67281498-67281498 on Build GRCh38

View Full Product Details