Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20737863-20738108 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THUMPD1
Entrez Gene ID
55623
Gene Name
THUMP domain containing 1
Gene Aliases
Tan1
UniGene
Hs.700004
Gene Chromosome Location
Chr.16: 20730255-20741964 on Build GRCh38
Transcripts
NM_001304550.1 NM_017736.4 XM_011545891.1 XM_017023433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5639760 c.469C>T p.R157* Chr.16: 20737894-20737894 on Build GRCh38
6861044 c.407-7_407-6insT p.? Chr.16: 20737962-20737963 on Build GRCh38
1678892 c.439G>A p.D147N Chr.16: 20737924-20737924 on Build GRCh38
6801518 c.423G>A p.V141V Chr.16: 20737940-20737940 on Build GRCh38
6838995 c.433C>T p.L145F Chr.16: 20737930-20737930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1056663863 Chr.16: 20738041-20738041 on Build GRCh38
rs766340150 Chr.16: 20737924-20737924 on Build GRCh38
rs1053038264 Chr.16: 20737935-20737935 on Build GRCh38
rs956102024 Chr.16: 20737910-20737910 on Build GRCh38
rs377037430 Chr.16: 20737992-20737992 on Build GRCh38
rs747641514 Chr.16: 20737915-20737915 on Build GRCh38
rs910889324 Chr.16: 20737963-20737965 on Build GRCh38
rs774413818 Chr.16: 20737942-20737942 on Build GRCh38
rs896627235 Chr.16: 20738052-20738052 on Build GRCh38
rs369169357 Chr.16: 20737987-20737987 on Build GRCh38
rs1039057816 Chr.16: 20738008-20738008 on Build GRCh38
rs747573450 Chr.16: 20737896-20737896 on Build GRCh38
rs536835370 Chr.16: 20738074-20738074 on Build GRCh38
rs889565816 Chr.16: 20738009-20738009 on Build GRCh38
rs776244641 Chr.16: 20737907-20737907 on Build GRCh38
rs769437686 Chr.16: 20737920-20737920 on Build GRCh38
rs368769699 Chr.16: 20737955-20737955 on Build GRCh38
rs922381253 Chr.16: 20737933-20737933 on Build GRCh38
rs201866042 Chr.16: 20737911-20737911 on Build GRCh38
rs930150218 Chr.16: 20738023-20738023 on Build GRCh38
rs141498187 Chr.16: 20737947-20737947 on Build GRCh38
rs1012344991 Chr.16: 20738055-20738055 on Build GRCh38
rs1011682244 Chr.16: 20737960-20737960 on Build GRCh38
rs757945159 Chr.16: 20737894-20737895 on Build GRCh38
rs768132475 Chr.16: 20737900-20737900 on Build GRCh38
rs772750661 Chr.16: 20737921-20737921 on Build GRCh38
rs754766868 Chr.16: 20737962-20737963 on Build GRCh38
rs779803167 Chr.16: 20737957-20737958 on Build GRCh38
rs751523219 Chr.16: 20737963-20737963 on Build GRCh38

View Full Product Details