Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 196596-196869 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP14
Entrez Gene ID
9097
Gene Name
ubiquitin specific peptidase 14
Gene Aliases
TGT
UniGene
Hs.464416
Gene Chromosome Location
Chr.18: 158483-213739 on Build GRCh38
Transcripts
NM_001037334.1 NM_005151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1387988 c.561C>T p.A187A Chr.18: 196734-196734 on Build GRCh38
6003333 c.589C>T p.Q197* Chr.18: 196762-196762 on Build GRCh38
4071461 c.560C>T p.A187V Chr.18: 196733-196733 on Build GRCh38
6082300 c.574C>A p.Q192K Chr.18: 196747-196747 on Build GRCh38
7309768 c.591A>G p.Q197Q Chr.18: 196764-196764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748053649 Chr.18: 196639-196639 on Build GRCh38
rs766313488 Chr.18: 196694-196694 on Build GRCh38
rs748883650 Chr.18: 196797-196797 on Build GRCh38
rs754593609 Chr.18: 196804-196804 on Build GRCh38
rs752722339 Chr.18: 196714-196714 on Build GRCh38
rs902328409 Chr.18: 196748-196748 on Build GRCh38
rs374980775 Chr.18: 196734-196734 on Build GRCh38
rs752525633 Chr.18: 196719-196719 on Build GRCh38
rs1010985674 Chr.18: 196736-196736 on Build GRCh38
rs764530928 Chr.18: 196740-196740 on Build GRCh38
rs966715416 Chr.18: 196837-196837 on Build GRCh38
rs199771859 Chr.18: 196777-196777 on Build GRCh38
rs111543418 Chr.18: 196732-196732 on Build GRCh38
rs773014517 Chr.18: 196679-196679 on Build GRCh38
rs960578042 Chr.18: 196827-196827 on Build GRCh38
rs370518955 Chr.18: 196696-196696 on Build GRCh38
rs759892412 Chr.18: 196698-196698 on Build GRCh38
rs865913135 Chr.18: 196733-196733 on Build GRCh38
rs752108484 Chr.18: 196754-196754 on Build GRCh38
rs1784381 Chr.18: 196657-196657 on Build GRCh38
rs771938055 Chr.18: 196668-196668 on Build GRCh38
rs572294024 Chr.18: 196713-196713 on Build GRCh38
rs757614604 Chr.18: 196757-196757 on Build GRCh38
rs655781 Chr.18: 196829-196829 on Build GRCh38

View Full Product Details