Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 246853000-246853225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AHCTF1
Entrez Gene ID
25909
Gene Name
AT-hook containing transcription factor 1
Gene Aliases
ELYS, MST108, MSTP108, TMBS62
UniGene
Hs.300887
Gene Chromosome Location
Chr.1: 246839098-246931978 on Build GRCh38
Transcripts
NM_001323342.1 NM_001323343.1 NM_015446.4 NR_136586.1 XM_006711758.1 XM_011544156.2 XM_011544157.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
383776 c.4537A>T p.S1513C Chr.1: 246853117-246853117 on Build GRCh38
140406 c.4489C>A p.L1497I Chr.1: 246853165-246853165 on Build GRCh38
6596557 c.4590+2T>C p.? Chr.1: 246853089-246853089 on Build GRCh38
4820233 c.4452G>A p.L1484L Chr.1: 246853202-246853202 on Build GRCh38
906892 c.4432C>A p.L1478I Chr.1: 246853222-246853222 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775050161 Chr.1: 246853040-246853040 on Build GRCh38
rs143330344 Chr.1: 246853205-246853205 on Build GRCh38
rs767807860 Chr.1: 246853168-246853168 on Build GRCh38
rs778634742 Chr.1: 246853185-246853185 on Build GRCh38
rs961319814 Chr.1: 246853144-246853144 on Build GRCh38
rs745817083 Chr.1: 246853190-246853190 on Build GRCh38
rs956913074 Chr.1: 246853092-246853092 on Build GRCh38
rs923551656 Chr.1: 246853156-246853156 on Build GRCh38
rs559480201 Chr.1: 246853029-246853029 on Build GRCh38
rs760179640 Chr.1: 246853051-246853051 on Build GRCh38
rs111792872 Chr.1: 246853057-246853057 on Build GRCh38
rs755618031 Chr.1: 246853080-246853081 on Build GRCh38
rs763611636 Chr.1: 246853110-246853110 on Build GRCh38
rs751633142 Chr.1: 246853129-246853129 on Build GRCh38
rs753359905 Chr.1: 246853093-246853094 on Build GRCh38
rs774160350 Chr.1: 246853064-246853066 on Build GRCh38
rs752692575 Chr.1: 246853081-246853081 on Build GRCh38
rs767000209 Chr.1: 246853111-246853111 on Build GRCh38
rs1029363366 Chr.1: 246853189-246853189 on Build GRCh38
rs755132680 Chr.1: 246853159-246853159 on Build GRCh38
rs12123173 Chr.1: 246853084-246853084 on Build GRCh38
rs976292015 Chr.1: 246853150-246853150 on Build GRCh38
rs776167403 Chr.1: 246853058-246853058 on Build GRCh38
rs750919421 Chr.1: 246853098-246853098 on Build GRCh38
rs756885510 Chr.1: 246853188-246853190 on Build GRCh38
rs765387503 Chr.1: 246853063-246853066 on Build GRCh38
rs931967771 Chr.1: 246853202-246853202 on Build GRCh38
rs1013084285 Chr.1: 246853071-246853073 on Build GRCh38
rs551508378 Chr.1: 246853059-246853059 on Build GRCh38
rs759293843 Chr.1: 246853070-246853071 on Build GRCh38
rs752906596 Chr.1: 246853170-246853170 on Build GRCh38
rs767470861 Chr.1: 246853074-246853075 on Build GRCh38
rs1015230590 Chr.1: 246853136-246853136 on Build GRCh38

View Full Product Details