Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 66150071-66150332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM55
Entrez Gene ID
84675
Gene Name
tripartite motif containing 55
Gene Aliases
MURF-2, RNF29, muRF2
UniGene
Hs.85524
Gene Chromosome Location
Chr.8: 66112667-66175485 on Build GRCh38
Transcripts
NM_033058.2 NM_184085.1 NM_184086.1 NM_184087.1 XM_005251316.4 XM_011517613.2 XM_011517614.2 XM_011517615.2 XM_011517616.2 XM_011517617.2 XM_017013908.1 XM_017013909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5767725 c.838-1G>A p.? Chr.8: 66150216-66150216 on Build GRCh38
5767726 c.838-1G>A p.? Chr.8: 66150216-66150216 on Build GRCh38
7296088 c.858A>T p.K286N Chr.8: 66150237-66150237 on Build GRCh38
7296087 c.858A>T p.K286N Chr.8: 66150237-66150237 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032046755 Chr.8: 66150165-66150165 on Build GRCh38
rs908736460 Chr.8: 66150268-66150268 on Build GRCh38
rs756204569 Chr.8: 66150308-66150308 on Build GRCh38
rs1012684091 Chr.8: 66150119-66150119 on Build GRCh38
rs775754774 Chr.8: 66150199-66150200 on Build GRCh38
rs373205100 Chr.8: 66150306-66150306 on Build GRCh38
rs753729648 Chr.8: 66150239-66150239 on Build GRCh38
rs953992152 Chr.8: 66150294-66150294 on Build GRCh38
rs755170441 Chr.8: 66150291-66150291 on Build GRCh38
rs751727697 Chr.8: 66150270-66150270 on Build GRCh38
rs770301512 Chr.8: 66150168-66150168 on Build GRCh38
rs767331404 Chr.8: 66150208-66150208 on Build GRCh38
rs1028173129 Chr.8: 66150252-66150252 on Build GRCh38
rs373060435 Chr.8: 66150191-66150191 on Build GRCh38
rs866140695 Chr.8: 66150124-66150124 on Build GRCh38
rs199933238 Chr.8: 66150250-66150250 on Build GRCh38
rs749777269 Chr.8: 66150306-66150307 on Build GRCh38
rs770184721 Chr.8: 66150194-66150195 on Build GRCh38
rs796691279 Chr.8: 66150105-66150105 on Build GRCh38
rs111259636 Chr.8: 66150134-66150134 on Build GRCh38
rs773658008 Chr.8: 66150170-66150170 on Build GRCh38
rs568848577 Chr.8: 66150108-66150108 on Build GRCh38
rs764082111 Chr.8: 66150234-66150234 on Build GRCh38
rs765043712 Chr.8: 66150253-66150253 on Build GRCh38
rs368967786 Chr.8: 66150173-66150173 on Build GRCh38
rs377022301 Chr.8: 66150214-66150214 on Build GRCh38
rs760172747 Chr.8: 66150228-66150228 on Build GRCh38
rs904196997 Chr.8: 66150136-66150136 on Build GRCh38
rs998515319 Chr.8: 66150156-66150156 on Build GRCh38
rs758579699 Chr.8: 66150206-66150206 on Build GRCh38
rs774106883 Chr.8: 66150200-66150200 on Build GRCh38
rs775112955 Chr.8: 66150227-66150227 on Build GRCh38
rs759284879 Chr.8: 66150202-66150202 on Build GRCh38

View Full Product Details