Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 151839188-151839443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNB4
Entrez Gene ID
785
Gene Name
calcium voltage-gated channel auxiliary subunit beta 4
Gene Aliases
CAB4, CACNLB4, EA5, EIG9, EJM, EJM4, EJM6
UniGene
Hs.120725
Gene Chromosome Location
Chr.2: 151832771-152099988 on Build GRCh38
Transcripts
NM_000726.3 NM_001005746.2 NM_001005747.2 NM_001145798.1 NM_001320722.1 XM_011511795.2 XM_011511796.2 XM_011511797.2 XM_017004880.1 XM_017004881.1 XM_017004882.1 XM_017004883.1 XM_017004884.1 XM_017004885.1 XM_017004886.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618669 c.1372T>A p.S458T Chr.2: 151839310-151839310 on Build GRCh38
7305637 c.1495G>A p.D499N Chr.2: 151839190-151839190 on Build GRCh38
4085626 c.1402G>A p.A468T Chr.2: 151839283-151839283 on Build GRCh38
4929395 c.1423T>A p.L475M Chr.2: 151839262-151839262 on Build GRCh38
5547634 c.1296_1305del10 p.A433fs*>51 Chr.2: 151839275-151839284 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771699585 Chr.2: 151839214-151839214 on Build GRCh38
rs143442080 Chr.2: 151839382-151839382 on Build GRCh38
rs763718335 Chr.2: 151839231-151839231 on Build GRCh38
rs754375350 Chr.2: 151839234-151839234 on Build GRCh38
rs750854413 Chr.2: 151839243-151839243 on Build GRCh38
rs746707937 Chr.2: 151839310-151839310 on Build GRCh38
rs368111233 Chr.2: 151839264-151839264 on Build GRCh38
rs766706127 Chr.2: 151839369-151839369 on Build GRCh38
rs529888359 Chr.2: 151839373-151839373 on Build GRCh38
rs775009719 Chr.2: 151839390-151839390 on Build GRCh38
rs765688685 Chr.2: 151839242-151839242 on Build GRCh38
rs763330914 Chr.2: 151839354-151839354 on Build GRCh38
rs375245807 Chr.2: 151839280-151839280 on Build GRCh38
rs779754184 Chr.2: 151839404-151839404 on Build GRCh38
rs757244766 Chr.2: 151839278-151839278 on Build GRCh38
rs749219941 Chr.2: 151839276-151839276 on Build GRCh38
rs745608627 Chr.2: 151839284-151839284 on Build GRCh38
rs760221470 Chr.2: 151839229-151839229 on Build GRCh38
rs794727118 Chr.2: 151839245-151839245 on Build GRCh38
rs758701126 Chr.2: 151839246-151839246 on Build GRCh38
rs1805032 Chr.2: 151839238-151839238 on Build GRCh38
rs775008250 Chr.2: 151839222-151839222 on Build GRCh38
rs796052291 Chr.2: 151839351-151839351 on Build GRCh38
rs925940329 Chr.2: 151839319-151839319 on Build GRCh38
rs751754026 Chr.2: 151839372-151839372 on Build GRCh38
rs773625599 Chr.2: 151839334-151839334 on Build GRCh38
rs796052292 Chr.2: 151839339-151839339 on Build GRCh38
rs755031838 Chr.2: 151839265-151839265 on Build GRCh38
rs1805029 Chr.2: 151839269-151839269 on Build GRCh38
rs941708614 Chr.2: 151839261-151839261 on Build GRCh38
rs768245741 Chr.2: 151839324-151839324 on Build GRCh38
rs771793003 Chr.2: 151839295-151839295 on Build GRCh38
rs758188877 Chr.2: 151839401-151839401 on Build GRCh38
rs766586063 Chr.2: 151839263-151839263 on Build GRCh38
rs979413676 Chr.2: 151839291-151839291 on Build GRCh38
rs762394421 Chr.2: 151839327-151839327 on Build GRCh38
rs761712022 Chr.2: 151839309-151839309 on Build GRCh38
rs768385119 Chr.2: 151839212-151839212 on Build GRCh38
rs778868244 Chr.2: 151839385-151839385 on Build GRCh38
rs745814577 Chr.2: 151839386-151839386 on Build GRCh38
rs762445852 Chr.2: 151839239-151839239 on Build GRCh38
rs752717348 Chr.2: 151839379-151839379 on Build GRCh38
rs765715570 Chr.2: 151839333-151839333 on Build GRCh38

View Full Product Details