Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31723826-31724079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf25
Entrez Gene ID
80739
Gene Name
chromosome 6 open reading frame 25
Gene Aliases
G6b, NG31
UniGene
Hs.247879
Gene Chromosome Location
Chr.6: 31720557-31726710 on Build GRCh38
Transcripts
NM_025260.3 NM_138272.2 NM_138273.2 NM_138274.2 NM_138275.2 NM_138277.2 XM_011514920.2 XM_011514921.2 XM_017011330.1 XM_017011331.1 XM_017011332.1 XM_017011333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4667914 c.353C>T p.T118I Chr.6: 31723930-31723930 on Build GRCh38
5021521 c.293T>C p.L98S Chr.6: 31723870-31723870 on Build GRCh38
7134560 c.334C>A p.H112N Chr.6: 31723911-31723911 on Build GRCh38
1077645 c.342C>T p.D114D Chr.6: 31723919-31723919 on Build GRCh38
7134559 c.334C>A p.H112N Chr.6: 31723911-31723911 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756580795 Chr.6: 31723944-31723944 on Build GRCh38
rs4947252 Chr.6: 31723889-31723889 on Build GRCh38
rs899727600 Chr.6: 31723935-31723935 on Build GRCh38
rs761490149 Chr.6: 31723979-31723979 on Build GRCh38
rs754708610 Chr.6: 31724025-31724025 on Build GRCh38
rs758095473 Chr.6: 31723852-31723852 on Build GRCh38
rs749638548 Chr.6: 31723949-31723949 on Build GRCh38
rs764525124 Chr.6: 31723860-31723861 on Build GRCh38
rs767576991 Chr.6: 31723892-31723892 on Build GRCh38
rs576980709 Chr.6: 31724049-31724049 on Build GRCh38
rs767118058 Chr.6: 31723988-31723988 on Build GRCh38
rs780701124 Chr.6: 31723940-31723940 on Build GRCh38
rs777620756 Chr.6: 31723854-31723854 on Build GRCh38
rs773187697 Chr.6: 31723894-31723894 on Build GRCh38
rs774612846 Chr.6: 31723887-31723887 on Build GRCh38
rs1056228711 Chr.6: 31724033-31724033 on Build GRCh38
rs779296363 Chr.6: 31724034-31724034 on Build GRCh38
rs775638872 Chr.6: 31723972-31723972 on Build GRCh38
rs749929656 Chr.6: 31723994-31723994 on Build GRCh38
rs200622244 Chr.6: 31723974-31723974 on Build GRCh38
rs34547016 Chr.6: 31723844-31723844 on Build GRCh38
rs758328832 Chr.6: 31723922-31723922 on Build GRCh38
rs955375841 Chr.6: 31723899-31723899 on Build GRCh38
rs752572699 Chr.6: 31723848-31723848 on Build GRCh38
rs1033898790 Chr.6: 31724059-31724059 on Build GRCh38
rs768034908 Chr.6: 31723930-31723931 on Build GRCh38
rs941524173 Chr.6: 31723928-31723928 on Build GRCh38
rs896130699 Chr.6: 31724039-31724039 on Build GRCh38
rs780585387 Chr.6: 31723946-31723946 on Build GRCh38
rs373460308 Chr.6: 31723976-31723976 on Build GRCh38
rs753518836 Chr.6: 31723967-31723967 on Build GRCh38
rs111732872 Chr.6: 31723989-31723989 on Build GRCh38
rs773313019 Chr.6: 31723956-31723956 on Build GRCh38
rs755497788 Chr.6: 31724025-31724025 on Build GRCh38
rs780536057 Chr.6: 31724000-31724000 on Build GRCh38
rs759479340 Chr.6: 31723913-31723913 on Build GRCh38
rs760718640 Chr.6: 31723961-31723961 on Build GRCh38
rs746324362 Chr.6: 31723879-31723879 on Build GRCh38
rs995443198 Chr.6: 31724024-31724024 on Build GRCh38
rs772216041 Chr.6: 31723955-31723955 on Build GRCh38
rs756862856 Chr.6: 31723874-31723874 on Build GRCh38
rs116400360 Chr.6: 31724021-31724021 on Build GRCh38
rs769075410 Chr.6: 31723950-31723950 on Build GRCh38
rs769843224 Chr.6: 31723969-31723969 on Build GRCh38
rs749691617 Chr.6: 31723886-31723886 on Build GRCh38
rs453098 Chr.6: 31723880-31723880 on Build GRCh38
rs752167162 Chr.6: 31723885-31723886 on Build GRCh38
rs369007488 Chr.6: 31723973-31723973 on Build GRCh38
rs748500146 Chr.6: 31724036-31724036 on Build GRCh38
rs765307090 Chr.6: 31723915-31723915 on Build GRCh38
rs763837376 Chr.6: 31723923-31723923 on Build GRCh38
rs765076070 Chr.6: 31723846-31723846 on Build GRCh38
rs757560451 Chr.6: 31723931-31723947 on Build GRCh38
rs751673323 Chr.6: 31723954-31723954 on Build GRCh38
rs1016040057 Chr.6: 31724019-31724019 on Build GRCh38
rs1054384537 Chr.6: 31723926-31723926 on Build GRCh38
rs756870049 Chr.6: 31723927-31723927 on Build GRCh38
rs749928985 Chr.6: 31723941-31723941 on Build GRCh38
rs779254034 Chr.6: 31723953-31723953 on Build GRCh38
rs775790329 Chr.6: 31723883-31723883 on Build GRCh38
rs771879584 Chr.6: 31723869-31723869 on Build GRCh38
rs182774419 Chr.6: 31723925-31723925 on Build GRCh38
rs752521425 Chr.6: 31723918-31723918 on Build GRCh38
rs755680998 Chr.6: 31723997-31723997 on Build GRCh38

View Full Product Details