Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 131581160-131581417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARG1
Entrez Gene ID
383
Gene Name
arginase 1
Gene Aliases
-
UniGene
Hs.440934
Gene Chromosome Location
Chr.6: 131573159-131584332 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Gene Symbol
MED23
Entrez Gene ID
9439
Gene Name
mediator complex subunit 23
Gene Aliases
ARC130, CRSP130, CRSP133, CRSP3, DRIP130, MRT18, SUR-2, SUR2
UniGene
Hs.29679
Gene Chromosome Location
Chr.6: 131573966-131628239 on Build GRCh38
Transcripts
NM_000045.3 NM_001244438.1 XM_011535801.2 NM_001270521.1 NM_015979.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7246112 c.355G>A p.G119R Chr.6: 131581268-131581268 on Build GRCh38
4662699 c.330C>T p.G110G Chr.6: 131581243-131581243 on Build GRCh38
2156578 c.306-4A>G p.? Chr.6: 131581215-131581215 on Build GRCh38
6602976 c.354C>T p.G118G Chr.6: 131581243-131581243 on Build GRCh38
4901955 c.380C>A p.T127N Chr.6: 131581293-131581293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750937576 Chr.6: 131581360-131581360 on Build GRCh38
rs775638241 Chr.6: 131581209-131581209 on Build GRCh38
rs747883555 Chr.6: 131581306-131581306 on Build GRCh38
rs765859028 Chr.6: 131581320-131581320 on Build GRCh38
rs762306280 Chr.6: 131581227-131581227 on Build GRCh38
rs104893947 Chr.6: 131581278-131581278 on Build GRCh38
rs766750593 Chr.6: 131581203-131581204 on Build GRCh38
rs913575060 Chr.6: 131581281-131581281 on Build GRCh38
rs749043086 Chr.6: 131581222-131581222 on Build GRCh38
rs927479708 Chr.6: 131581376-131581376 on Build GRCh38
rs771165013 Chr.6: 131581299-131581299 on Build GRCh38
rs765617697 Chr.6: 131581346-131581346 on Build GRCh38
rs774535167 Chr.6: 131581303-131581303 on Build GRCh38
rs745875961 Chr.6: 131581197-131581197 on Build GRCh38
rs747268449 Chr.6: 131581289-131581289 on Build GRCh38
rs751231138 Chr.6: 131581255-131581255 on Build GRCh38
rs767219084 Chr.6: 131581338-131581338 on Build GRCh38
rs754259099 Chr.6: 131581341-131581341 on Build GRCh38
rs746381302 Chr.6: 131581275-131581275 on Build GRCh38
rs200458963 Chr.6: 131581246-131581246 on Build GRCh38
rs985914059 Chr.6: 131581204-131581204 on Build GRCh38
rs762633715 Chr.6: 131581328-131581329 on Build GRCh38
rs769778089 Chr.6: 131581310-131581310 on Build GRCh38
rs780157621 Chr.6: 131581280-131581280 on Build GRCh38
rs758905397 Chr.6: 131581379-131581379 on Build GRCh38
rs776376445 Chr.6: 131581215-131581215 on Build GRCh38
rs754205928 Chr.6: 131581264-131581264 on Build GRCh38
rs41285336 Chr.6: 131581189-131581189 on Build GRCh38
rs772341418 Chr.6: 131581203-131581203 on Build GRCh38
rs535456448 Chr.6: 131581319-131581319 on Build GRCh38
rs757852473 Chr.6: 131581266-131581266 on Build GRCh38
rs777553747 Chr.6: 131581214-131581214 on Build GRCh38
rs574320679 Chr.6: 131581305-131581305 on Build GRCh38
rs946323935 Chr.6: 131581330-131581330 on Build GRCh38
rs757762475 Chr.6: 131581342-131581342 on Build GRCh38
rs776939220 Chr.6: 131581284-131581285 on Build GRCh38
rs779072486 Chr.6: 131581196-131581196 on Build GRCh38
rs754655577 Chr.6: 131581262-131581262 on Build GRCh38
rs749966465 Chr.6: 131581226-131581226 on Build GRCh38
rs866733989 Chr.6: 131581285-131581285 on Build GRCh38
rs537503803 Chr.6: 131581212-131581212 on Build GRCh38
rs761773853 Chr.6: 131581221-131581221 on Build GRCh38
rs779219793 Chr.6: 131581273-131581273 on Build GRCh38
rs140549609 Chr.6: 131581296-131581296 on Build GRCh38
rs995938048 Chr.6: 131581225-131581225 on Build GRCh38
rs759282838 Chr.6: 131581324-131581324 on Build GRCh38
rs543245025 Chr.6: 131581200-131581200 on Build GRCh38
rs104893943 Chr.6: 131581326-131581326 on Build GRCh38
rs763780314 Chr.6: 131581388-131581389 on Build GRCh38
rs773968554 Chr.6: 131581321-131581321 on Build GRCh38

View Full Product Details