Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2407180-2407411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRPM5
Entrez Gene ID
29850
Gene Name
transient receptor potential cation channel subfamily M member 5
Gene Aliases
LTRPC5, MTR1
UniGene
Hs.272287
Gene Chromosome Location
Chr.11: 2403962-2423424 on Build GRCh38
Transcripts
NM_014555.3 XM_017017628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5485433 c.3046C>T p.L1016L Chr.11: 2407191-2407191 on Build GRCh38
2112823 c.3033C>T p.A1011A Chr.11: 2407204-2407204 on Build GRCh38
3967462 c.2942C>T p.T981M Chr.11: 2407295-2407295 on Build GRCh38
6809576 c.2994C>T p.Y998Y Chr.11: 2407243-2407243 on Build GRCh38
3676147 c.3036G>T p.P1012P Chr.11: 2407201-2407201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038501220 Chr.11: 2407232-2407232 on Build GRCh38
rs200136506 Chr.11: 2407247-2407247 on Build GRCh38
rs369160570 Chr.11: 2407341-2407341 on Build GRCh38
rs760152144 Chr.11: 2407213-2407213 on Build GRCh38
rs369338949 Chr.11: 2407322-2407322 on Build GRCh38
rs930177825 Chr.11: 2407384-2407384 on Build GRCh38
rs754650023 Chr.11: 2407261-2407261 on Build GRCh38
rs762115012 Chr.11: 2407295-2407295 on Build GRCh38
rs750309104 Chr.11: 2407239-2407239 on Build GRCh38
rs773280467 Chr.11: 2407308-2407308 on Build GRCh38
rs146751651 Chr.11: 2407218-2407218 on Build GRCh38
rs369339407 Chr.11: 2407204-2407204 on Build GRCh38
rs751636093 Chr.11: 2407221-2407221 on Build GRCh38
rs372751105 Chr.11: 2407326-2407326 on Build GRCh38
rs199741498 Chr.11: 2407265-2407265 on Build GRCh38
rs768890047 Chr.11: 2407290-2407290 on Build GRCh38
rs149842796 Chr.11: 2407332-2407332 on Build GRCh38
rs150296155 Chr.11: 2407222-2407222 on Build GRCh38
rs774395803 Chr.11: 2407294-2407294 on Build GRCh38
rs555023607 Chr.11: 2407356-2407356 on Build GRCh38
rs753704388 Chr.11: 2407248-2407248 on Build GRCh38
rs201364238 Chr.11: 2407331-2407331 on Build GRCh38
rs747674181 Chr.11: 2407205-2407205 on Build GRCh38
rs998733668 Chr.11: 2407343-2407343 on Build GRCh38
rs770236520 Chr.11: 2407215-2407215 on Build GRCh38
rs773538776 Chr.11: 2407357-2407357 on Build GRCh38
rs760598324 Chr.11: 2407314-2407314 on Build GRCh38
rs984325837 Chr.11: 2407361-2407361 on Build GRCh38
rs140407752 Chr.11: 2407219-2407219 on Build GRCh38
rs753611763 Chr.11: 2407321-2407321 on Build GRCh38
rs746585320 Chr.11: 2407342-2407342 on Build GRCh38
rs766365530 Chr.11: 2407317-2407317 on Build GRCh38
rs371930632 Chr.11: 2407266-2407266 on Build GRCh38
rs748877715 Chr.11: 2407201-2407201 on Build GRCh38
rs375833516 Chr.11: 2407217-2407217 on Build GRCh38
rs1023748956 Chr.11: 2407311-2407311 on Build GRCh38
rs533590172 Chr.11: 2407351-2407351 on Build GRCh38
rs756055517 Chr.11: 2407241-2407241 on Build GRCh38
rs768298281 Chr.11: 2407202-2407202 on Build GRCh38
rs771449920 Chr.11: 2407208-2407208 on Build GRCh38
rs529067735 Chr.11: 2407272-2407272 on Build GRCh38
rs749841634 Chr.11: 2407350-2407350 on Build GRCh38
rs911263358 Chr.11: 2407336-2407336 on Build GRCh38
rs908917087 Chr.11: 2407368-2407368 on Build GRCh38
rs550516385 Chr.11: 2407282-2407282 on Build GRCh38
rs58738928 Chr.11: 2407303-2407303 on Build GRCh38
rs749718995 Chr.11: 2407288-2407288 on Build GRCh38
rs770430536 Chr.11: 2407273-2407273 on Build GRCh38
rs780747022 Chr.11: 2407348-2407348 on Build GRCh38
rs777047940 Chr.11: 2407212-2407212 on Build GRCh38
rs756665554 Chr.11: 2407345-2407345 on Build GRCh38
rs778481579 Chr.11: 2407264-2407264 on Build GRCh38

View Full Product Details