Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 36341752-36342009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK40
Entrez Gene ID
83931
Gene Name
serine/threonine kinase 40
Gene Aliases
SHIK, SgK495
UniGene
Hs.471768
Gene Chromosome Location
Chr.1: 36339619-36385927 on Build GRCh38
Transcripts
NM_001282546.1 NM_001282547.1 NM_032017.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507186 c.1258A>T p.M420L Chr.1: 36341805-36341805 on Build GRCh38
6497134 c.1314G>C p.L438L Chr.1: 36341764-36341764 on Build GRCh38
142972 c.1256_1257CC>TT p.P419L Chr.1: 36341806-36341807 on Build GRCh38
4237378 c.1199C>T p.A400V Chr.1: 36341879-36341879 on Build GRCh38
330292 c.1183G>A p.A395T Chr.1: 36341880-36341880 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925140408 Chr.1: 36341819-36341819 on Build GRCh38
rs759242925 Chr.1: 36341814-36341814 on Build GRCh38
rs780619377 Chr.1: 36341772-36341772 on Build GRCh38
rs3795498 Chr.1: 36341880-36341880 on Build GRCh38
rs755430415 Chr.1: 36341890-36341890 on Build GRCh38
rs762527980 Chr.1: 36341791-36341791 on Build GRCh38
rs769273778 Chr.1: 36341789-36341789 on Build GRCh38
rs373009034 Chr.1: 36341905-36341905 on Build GRCh38
rs146560631 Chr.1: 36341971-36341971 on Build GRCh38
rs201662493 Chr.1: 36341836-36341836 on Build GRCh38
rs556999586 Chr.1: 36341978-36341978 on Build GRCh38
rs754087375 Chr.1: 36341817-36341817 on Build GRCh38
rs202225847 Chr.1: 36341963-36341963 on Build GRCh38
rs199625181 Chr.1: 36341877-36341877 on Build GRCh38
rs564284695 Chr.1: 36341844-36341844 on Build GRCh38
rs780818464 Chr.1: 36341777-36341777 on Build GRCh38
rs747283676 Chr.1: 36341774-36341774 on Build GRCh38
rs745399557 Chr.1: 36341932-36341932 on Build GRCh38
rs574330868 Chr.1: 36341790-36341790 on Build GRCh38
rs3795499 Chr.1: 36341851-36341851 on Build GRCh38
rs770162943 Chr.1: 36341973-36341973 on Build GRCh38
rs772070192 Chr.1: 36341942-36341942 on Build GRCh38
rs753269282 Chr.1: 36341982-36341982 on Build GRCh38
rs56405327 Chr.1: 36341962-36341962 on Build GRCh38
rs781415766 Chr.1: 36341889-36341889 on Build GRCh38
rs774376244 Chr.1: 36341972-36341972 on Build GRCh38
rs970241157 Chr.1: 36341885-36341885 on Build GRCh38
rs139437228 Chr.1: 36341876-36341876 on Build GRCh38
rs200986219 Chr.1: 36341904-36341904 on Build GRCh38
rs202016833 Chr.1: 36341893-36341893 on Build GRCh38
rs143287658 Chr.1: 36341776-36341776 on Build GRCh38
rs55926307 Chr.1: 36341899-36341899 on Build GRCh38
rs753687568 Chr.1: 36341829-36341829 on Build GRCh38
rs768014184 Chr.1: 36341891-36341891 on Build GRCh38
rs755305561 Chr.1: 36341843-36341843 on Build GRCh38
rs201445970 Chr.1: 36341845-36341845 on Build GRCh38
rs768567087 Chr.1: 36341948-36341948 on Build GRCh38
rs1055733693 Chr.1: 36341921-36341921 on Build GRCh38
rs767997763 Chr.1: 36341981-36341981 on Build GRCh38
rs752175420 Chr.1: 36341837-36341837 on Build GRCh38
rs778434947 Chr.1: 36341924-36341924 on Build GRCh38
rs756696476 Chr.1: 36341907-36341907 on Build GRCh38
rs752154301 Chr.1: 36341962-36341963 on Build GRCh38
rs774405030 Chr.1: 36341801-36341801 on Build GRCh38
rs377262836 Chr.1: 36341821-36341821 on Build GRCh38
rs775585540 Chr.1: 36341943-36341943 on Build GRCh38
rs747840660 Chr.1: 36341788-36341788 on Build GRCh38
rs575657248 Chr.1: 36341828-36341828 on Build GRCh38
rs530029776 Chr.1: 36341856-36341856 on Build GRCh38
rs148851878 Chr.1: 36341863-36341863 on Build GRCh38
rs754172776 Chr.1: 36341986-36341986 on Build GRCh38
rs917962796 Chr.1: 36341987-36341987 on Build GRCh38
rs756435731 Chr.1: 36341983-36341983 on Build GRCh38
rs765322359 Chr.1: 36341865-36341865 on Build GRCh38
rs200223642 Chr.1: 36341944-36341944 on Build GRCh38
rs758522274 Chr.1: 36341831-36341831 on Build GRCh38
rs996025547 Chr.1: 36341868-36341868 on Build GRCh38
rs770715643 Chr.1: 36341795-36341795 on Build GRCh38
rs777493861 Chr.1: 36341847-36341847 on Build GRCh38
rs112242536 Chr.1: 36341815-36341815 on Build GRCh38
rs116789558 Chr.1: 36341984-36341984 on Build GRCh38
rs200773119 Chr.1: 36341850-36341850 on Build GRCh38
rs775387498 Chr.1: 36341859-36341859 on Build GRCh38
rs759593248 Chr.1: 36341858-36341858 on Build GRCh38
rs866820712 Chr.1: 36341787-36341787 on Build GRCh38
rs557283950 Chr.1: 36341818-36341818 on Build GRCh38

View Full Product Details