Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 155506895-155507142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMLHE
Entrez Gene ID
55217
Gene Name
trimethyllysine hydroxylase, epsilon
Gene Aliases
AUTSX6, BBOX2, TMLD, TMLH, TMLHED, XAP130
UniGene
Hs.133321
Gene Chromosome Location
Chr.X: 155489007-155612961 on Build GRCh38
Transcripts
NM_001184797.1 NM_018196.3 XM_011531182.2

SNP Detail

SNP ID Chromosome Location
rs782584564 Chr.X: 155507098-155507098 on Build GRCh38
rs782682224 Chr.X: 155507002-155507002 on Build GRCh38
rs782255101 Chr.X: 155506939-155506939 on Build GRCh38
rs782177151 Chr.X: 155507108-155507108 on Build GRCh38
rs782751920 Chr.X: 155507081-155507081 on Build GRCh38
rs919633930 Chr.X: 155507071-155507071 on Build GRCh38
rs932846114 Chr.X: 155507079-155507079 on Build GRCh38
rs200416262 Chr.X: 155507034-155507034 on Build GRCh38
rs782537274 Chr.X: 155507095-155507095 on Build GRCh38
rs782803874 Chr.X: 155507051-155507051 on Build GRCh38
rs869320768 Chr.X: 155506931-155506932 on Build GRCh38
rs781990669 Chr.X: 155507037-155507037 on Build GRCh38
rs782171662 Chr.X: 155507045-155507045 on Build GRCh38
rs782372493 Chr.X: 155506993-155506993 on Build GRCh38
rs782532251 Chr.X: 155507067-155507067 on Build GRCh38
rs199989202 Chr.X: 155507053-155507053 on Build GRCh38
rs782643746 Chr.X: 155506930-155506930 on Build GRCh38
rs141804314 Chr.X: 155507027-155507027 on Build GRCh38
rs181549466 Chr.X: 155507093-155507093 on Build GRCh38
rs781824393 Chr.X: 155507065-155507065 on Build GRCh38
rs782703460 Chr.X: 155507015-155507015 on Build GRCh38
rs782279353 Chr.X: 155507011-155507011 on Build GRCh38
rs782222839 Chr.X: 155507032-155507032 on Build GRCh38
rs782761175 Chr.X: 155507054-155507054 on Build GRCh38
rs782793284 Chr.X: 155506956-155506956 on Build GRCh38

View Full Product Details