Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 107753356-107753615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBLL1
Entrez Gene ID
79872
Gene Name
Cbl proto-oncogene like 1
Gene Aliases
HAKAI, RNF188
UniGene
Hs.592271
Gene Chromosome Location
Chr.7: 107743497-107764997 on Build GRCh38
Transcripts
NM_001284291.1 NM_024814.3 NR_024199.2 XM_011516580.2 XM_017012643.1 XM_017012644.1 XM_017012645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6295153 c.210G>T p.R70R Chr.7: 107753439-107753439 on Build GRCh38
4836683 c.202G>C p.E68Q Chr.7: 107753431-107753431 on Build GRCh38
3631792 c.268T>A p.F90I Chr.7: 107753497-107753497 on Build GRCh38
7210355 c.241A>G p.N81D Chr.7: 107753470-107753470 on Build GRCh38
3631791 c.256C>T p.P86S Chr.7: 107753485-107753485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368266123 Chr.7: 107753395-107753395 on Build GRCh38
rs754365005 Chr.7: 107753528-107753528 on Build GRCh38
rs576560556 Chr.7: 107753553-107753553 on Build GRCh38
rs746716765 Chr.7: 107753538-107753539 on Build GRCh38
rs766048496 Chr.7: 107753498-107753498 on Build GRCh38
rs760462656 Chr.7: 107753470-107753470 on Build GRCh38
rs1030754000 Chr.7: 107753403-107753403 on Build GRCh38
rs746147506 Chr.7: 107753437-107753437 on Build GRCh38
rs536454381 Chr.7: 107753453-107753453 on Build GRCh38
rs751173605 Chr.7: 107753521-107753521 on Build GRCh38
rs375230713 Chr.7: 107753393-107753393 on Build GRCh38
rs766984678 Chr.7: 107753524-107753524 on Build GRCh38
rs750783671 Chr.7: 107753551-107753551 on Build GRCh38
rs757697015 Chr.7: 107753426-107753426 on Build GRCh38
rs779466531 Chr.7: 107753550-107753550 on Build GRCh38
rs780225140 Chr.7: 107753447-107753447 on Build GRCh38
rs867094880 Chr.7: 107753428-107753428 on Build GRCh38
rs958330623 Chr.7: 107753391-107753391 on Build GRCh38
rs140334387 Chr.7: 107753423-107753423 on Build GRCh38
rs867537475 Chr.7: 107753570-107753570 on Build GRCh38
rs747093711 Chr.7: 107753452-107753452 on Build GRCh38
rs762644254 Chr.7: 107753493-107753493 on Build GRCh38
rs780815918 Chr.7: 107753564-107753572 on Build GRCh38
rs754470430 Chr.7: 107753408-107753408 on Build GRCh38
rs201049549 Chr.7: 107753411-107753411 on Build GRCh38
rs556313861 Chr.7: 107753477-107753477 on Build GRCh38
rs754348499 Chr.7: 107753428-107753430 on Build GRCh38
rs758980708 Chr.7: 107753523-107753523 on Build GRCh38
rs567570829 Chr.7: 107753438-107753438 on Build GRCh38
rs769644687 Chr.7: 107753475-107753475 on Build GRCh38
rs779412576 Chr.7: 107753427-107753427 on Build GRCh38
rs756700452 Chr.7: 107753539-107753540 on Build GRCh38
rs757821830 Chr.7: 107753544-107753544 on Build GRCh38
rs758008942 Chr.7: 107753397-107753397 on Build GRCh38

View Full Product Details