Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 56382906-56383139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AURKA
Entrez Gene ID
6790
Gene Name
aurora kinase A
Gene Aliases
AIK, ARK1, AURA, BTAK, PPP1R47, STK15, STK6, STK7
UniGene
Hs.250822
Gene Chromosome Location
Chr.20: 56369389-56392337 on Build GRCh38
Transcripts
NM_001323303.1 NM_001323304.1 NM_001323305.1 NM_003600.3 NM_198433.2 NM_198434.2 NM_198435.2 NM_198436.2 NM_198437.2 XM_017028034.1 XM_017028035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6972534 c.566+1G>A p.? Chr.20: 56382984-56382984 on Build GRCh38
6965176 c.518G>A p.G173E Chr.20: 56383033-56383033 on Build GRCh38
5554787 c.517G>A p.G173R Chr.20: 56383034-56383034 on Build GRCh38
5977603 c.460_461delCA p.Q154fs*8 Chr.20: 56383090-56383091 on Build GRCh38
5951932 c.516C>T p.A172A Chr.20: 56383035-56383035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769430412 Chr.20: 56383052-56383052 on Build GRCh38
rs948288770 Chr.20: 56383018-56383018 on Build GRCh38
rs998475564 Chr.20: 56382941-56382941 on Build GRCh38
rs143926563 Chr.20: 56383026-56383026 on Build GRCh38
rs746609481 Chr.20: 56383024-56383024 on Build GRCh38
rs45520831 Chr.20: 56383015-56383015 on Build GRCh38
rs529781803 Chr.20: 56382930-56382930 on Build GRCh38
rs972853385 Chr.20: 56382929-56382929 on Build GRCh38
rs377380364 Chr.20: 56383089-56383089 on Build GRCh38
rs200449915 Chr.20: 56382967-56382967 on Build GRCh38
rs148425454 Chr.20: 56383035-56383035 on Build GRCh38
rs771707391 Chr.20: 56382985-56382985 on Build GRCh38
rs960802070 Chr.20: 56382945-56382945 on Build GRCh38
rs992605483 Chr.20: 56382966-56382966 on Build GRCh38
rs763570140 Chr.20: 56383076-56383076 on Build GRCh38
rs751997979 Chr.20: 56383090-56383090 on Build GRCh38
rs752827659 Chr.20: 56382934-56382934 on Build GRCh38
rs201174158 Chr.20: 56382962-56382962 on Build GRCh38
rs773531333 Chr.20: 56383059-56383059 on Build GRCh38
rs370354724 Chr.20: 56382949-56382949 on Build GRCh38
rs766650881 Chr.20: 56383079-56383079 on Build GRCh38
rs779964295 Chr.20: 56382986-56382986 on Build GRCh38
rs1052598989 Chr.20: 56383104-56383104 on Build GRCh38
rs370497957 Chr.20: 56383034-56383034 on Build GRCh38
rs759659726 Chr.20: 56383103-56383103 on Build GRCh38

View Full Product Details