Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151881235-151881468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADACL2-AS1
Entrez Gene ID
101928142
Gene Name
AADACL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.680207
Gene Chromosome Location
Chr.3: 151751179-151928175 on Build GRCh38
Transcripts
NR_110202.1 NR_110203.1 NM_033050.5

Gene Symbol
SUCNR1
Entrez Gene ID
56670
Gene Name
succinate receptor 1
Gene Aliases
GPR91
UniGene
Hs.133181
Gene Chromosome Location
Chr.3: 151873643-151884619 on Build GRCh38
Transcripts
NR_110202.1 NR_110203.1 NM_033050.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3003902 c.830A>T p.Y277F Chr.3: 151881373-151881373 on Build GRCh38
6792792 c.771T>C p.A257A Chr.3: 151881314-151881314 on Build GRCh38
6485393 c.884A>G p.Y295C Chr.3: 151881427-151881427 on Build GRCh38
6511669 c.833T>A p.I278N Chr.3: 151881376-151881376 on Build GRCh38
3003898 c.775C>T p.R259C Chr.3: 151881318-151881318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768908503 Chr.3: 151881418-151881418 on Build GRCh38
rs199755765 Chr.3: 151881384-151881384 on Build GRCh38
rs200218473 Chr.3: 151881293-151881293 on Build GRCh38
rs146349417 Chr.3: 151881307-151881307 on Build GRCh38
rs373530839 Chr.3: 151881352-151881352 on Build GRCh38
rs772022838 Chr.3: 151881410-151881410 on Build GRCh38
rs766683190 Chr.3: 151881373-151881373 on Build GRCh38
rs776750149 Chr.3: 151881341-151881341 on Build GRCh38
rs201056449 Chr.3: 151881427-151881427 on Build GRCh38
rs757209317 Chr.3: 151881387-151881387 on Build GRCh38
rs774168564 Chr.3: 151881256-151881256 on Build GRCh38
rs148565296 Chr.3: 151881360-151881360 on Build GRCh38
rs747738623 Chr.3: 151881297-151881297 on Build GRCh38
rs140738447 Chr.3: 151881356-151881356 on Build GRCh38
rs760938730 Chr.3: 151881332-151881332 on Build GRCh38
rs375799840 Chr.3: 151881284-151881284 on Build GRCh38
rs557998857 Chr.3: 151881375-151881375 on Build GRCh38
rs778527962 Chr.3: 151881291-151881291 on Build GRCh38
rs374436394 Chr.3: 151881262-151881262 on Build GRCh38
rs775869786 Chr.3: 151881329-151881329 on Build GRCh38
rs142256005 Chr.3: 151881298-151881298 on Build GRCh38
rs951732383 Chr.3: 151881289-151881289 on Build GRCh38
rs780824530 Chr.3: 151881295-151881295 on Build GRCh38
rs767839041 Chr.3: 151881260-151881260 on Build GRCh38
rs771928792 Chr.3: 151881326-151881326 on Build GRCh38
rs749227853 Chr.3: 151881312-151881312 on Build GRCh38
rs1053762952 Chr.3: 151881417-151881417 on Build GRCh38
rs1045709223 Chr.3: 151881376-151881376 on Build GRCh38
rs556799089 Chr.3: 151881388-151881388 on Build GRCh38
rs760842906 Chr.3: 151881264-151881264 on Build GRCh38
rs947382915 Chr.3: 151881357-151881357 on Build GRCh38
rs760741154 Chr.3: 151881354-151881356 on Build GRCh38
rs745654334 Chr.3: 151881323-151881323 on Build GRCh38
rs570372662 Chr.3: 151881257-151881257 on Build GRCh38
rs142852744 Chr.3: 151881385-151881385 on Build GRCh38
rs915963264 Chr.3: 151881369-151881369 on Build GRCh38
rs779961865 Chr.3: 151881411-151881411 on Build GRCh38
rs768657266 Chr.3: 151881339-151881339 on Build GRCh38
rs201481949 Chr.3: 151881290-151881290 on Build GRCh38
rs770883727 Chr.3: 151881319-151881319 on Build GRCh38
rs747284179 Chr.3: 151881412-151881412 on Build GRCh38
rs372834017 Chr.3: 151881268-151881268 on Build GRCh38
rs368899419 Chr.3: 151881321-151881321 on Build GRCh38
rs912470352 Chr.3: 151881303-151881303 on Build GRCh38
rs957761712 Chr.3: 151881266-151881266 on Build GRCh38
rs201669822 Chr.3: 151881427-151881428 on Build GRCh38
rs745744082 Chr.3: 151881399-151881399 on Build GRCh38

View Full Product Details