Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 128573517-128573766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01565
Entrez Gene ID
23434
Gene Name
long intergenic non-protein coding RNA 1565
Gene Aliases
C3orf27, GR6
UniGene
Hs.194283
Gene Chromosome Location
Chr.3: 128572000-128576086 on Build GRCh38
Transcripts
NR_125802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4918708 c.84C>T p.R28R Chr.3: 128573646-128573646 on Build GRCh38
4979255 c.125G>A p.G42D Chr.3: 128573605-128573605 on Build GRCh38
4956427 c.56C>T p.T19M Chr.3: 128573674-128573674 on Build GRCh38
4529030 c.157G>A p.G53R Chr.3: 128573573-128573573 on Build GRCh38
4514023 c.95C>T p.P32L Chr.3: 128573635-128573635 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971566891 Chr.3: 128573590-128573590 on Build GRCh38
rs780392904 Chr.3: 128573742-128573742 on Build GRCh38
rs761811536 Chr.3: 128573699-128573699 on Build GRCh38
rs543666210 Chr.3: 128573655-128573655 on Build GRCh38
rs747490907 Chr.3: 128573606-128573606 on Build GRCh38
rs368941128 Chr.3: 128573580-128573580 on Build GRCh38
rs753756788 Chr.3: 128573705-128573705 on Build GRCh38
rs774453960 Chr.3: 128573634-128573634 on Build GRCh38
rs752761775 Chr.3: 128573535-128573535 on Build GRCh38
rs746201455 Chr.3: 128573679-128573679 on Build GRCh38
rs754668014 Chr.3: 128573709-128573709 on Build GRCh38
rs764135367 Chr.3: 128573632-128573632 on Build GRCh38
rs778566199 Chr.3: 128573666-128573666 on Build GRCh38
rs749481004 Chr.3: 128573689-128573689 on Build GRCh38
rs141693917 Chr.3: 128573654-128573654 on Build GRCh38
rs150531956 Chr.3: 128573674-128573674 on Build GRCh38
rs754721107 Chr.3: 128573664-128573664 on Build GRCh38
rs141391181 Chr.3: 128573545-128573545 on Build GRCh38
rs147972080 Chr.3: 128573610-128573610 on Build GRCh38
rs973115267 Chr.3: 128573728-128573728 on Build GRCh38
rs200800269 Chr.3: 128573700-128573700 on Build GRCh38
rs749123796 Chr.3: 128573708-128573708 on Build GRCh38
rs35480236 Chr.3: 128573640-128573640 on Build GRCh38
rs760716188 Chr.3: 128573704-128573704 on Build GRCh38
rs771412688 Chr.3: 128573607-128573607 on Build GRCh38
rs77910046 Chr.3: 128573605-128573605 on Build GRCh38
rs775716901 Chr.3: 128573630-128573630 on Build GRCh38
rs779578279 Chr.3: 128573581-128573581 on Build GRCh38
rs752190728 Chr.3: 128573715-128573715 on Build GRCh38
rs201887104 Chr.3: 128573550-128573550 on Build GRCh38
rs769988720 Chr.3: 128573614-128573614 on Build GRCh38
rs770073747 Chr.3: 128573682-128573682 on Build GRCh38
rs914571934 Chr.3: 128573676-128573676 on Build GRCh38
rs762967641 Chr.3: 128573631-128573631 on Build GRCh38
rs761732903 Chr.3: 128573636-128573636 on Build GRCh38
rs146933691 Chr.3: 128573582-128573582 on Build GRCh38
rs750869087 Chr.3: 128573719-128573719 on Build GRCh38
rs376936146 Chr.3: 128573718-128573718 on Build GRCh38
rs754557571 Chr.3: 128573602-128573602 on Build GRCh38
rs376175484 Chr.3: 128573733-128573733 on Build GRCh38
rs199850610 Chr.3: 128573684-128573684 on Build GRCh38
rs768932612 Chr.3: 128573690-128573690 on Build GRCh38
rs561186025 Chr.3: 128573576-128573576 on Build GRCh38
rs763022689 Chr.3: 128573537-128573537 on Build GRCh38
rs373793796 Chr.3: 128573611-128573611 on Build GRCh38
rs572324211 Chr.3: 128573549-128573549 on Build GRCh38
rs531686906 Chr.3: 128573594-128573594 on Build GRCh38
rs375844283 Chr.3: 128573647-128573647 on Build GRCh38
rs138294249 Chr.3: 128573698-128573698 on Build GRCh38
rs767411930 Chr.3: 128573640-128573640 on Build GRCh38
rs750249426 Chr.3: 128573570-128573570 on Build GRCh38
rs537931122 Chr.3: 128573724-128573724 on Build GRCh38
rs149005096 Chr.3: 128573539-128573539 on Build GRCh38
rs943530001 Chr.3: 128573545-128573545 on Build GRCh38
rs981568423 Chr.3: 128573624-128573624 on Build GRCh38
rs764998108 Chr.3: 128573712-128573712 on Build GRCh38
rs755894773 Chr.3: 128573648-128573648 on Build GRCh38
rs377636596 Chr.3: 128573673-128573673 on Build GRCh38

View Full Product Details