Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92340612-92340863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MARCH5
Entrez Gene ID
54708
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH-V, MITOL, RNF153
UniGene
Hs.573490
Gene Chromosome Location
Chr.10: 92291163-92353964 on Build GRCh38
Transcripts
NM_017824.4 XM_011539886.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1969573 c.268G>C p.D90H Chr.10: 92340702-92340702 on Build GRCh38
5640970 c.333G>T p.W111C Chr.10: 92340767-92340767 on Build GRCh38
4165571 c.319G>A p.G107S Chr.10: 92340753-92340753 on Build GRCh38
6130849 c.361G>T p.V121L Chr.10: 92340795-92340795 on Build GRCh38
5492676 c.253G>A p.V85I Chr.10: 92340687-92340687 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367545805 Chr.10: 92340704-92340704 on Build GRCh38
rs561390861 Chr.10: 92340670-92340670 on Build GRCh38
rs759741174 Chr.10: 92340688-92340688 on Build GRCh38
rs756421587 Chr.10: 92340823-92340823 on Build GRCh38
rs61760883 Chr.10: 92340677-92340677 on Build GRCh38
rs189096816 Chr.10: 92340687-92340687 on Build GRCh38
rs773736742 Chr.10: 92340811-92340811 on Build GRCh38
rs781468500 Chr.10: 92340653-92340653 on Build GRCh38
rs753879708 Chr.10: 92340713-92340713 on Build GRCh38
rs921963931 Chr.10: 92340661-92340662 on Build GRCh38
rs767763363 Chr.10: 92340691-92340691 on Build GRCh38
rs748486627 Chr.10: 92340659-92340659 on Build GRCh38
rs36076760 Chr.10: 92340782-92340782 on Build GRCh38
rs371622719 Chr.10: 92340734-92340734 on Build GRCh38
rs757230888 Chr.10: 92340728-92340728 on Build GRCh38
rs201912277 Chr.10: 92340703-92340703 on Build GRCh38
rs199682466 Chr.10: 92340669-92340669 on Build GRCh38
rs748826990 Chr.10: 92340652-92340654 on Build GRCh38
rs749938052 Chr.10: 92340746-92340746 on Build GRCh38
rs943832285 Chr.10: 92340744-92340744 on Build GRCh38
rs36004450 Chr.10: 92340686-92340686 on Build GRCh38
rs748542270 Chr.10: 92340816-92340816 on Build GRCh38
rs181021185 Chr.10: 92340707-92340707 on Build GRCh38
rs754653843 Chr.10: 92340812-92340812 on Build GRCh38
rs41313479 Chr.10: 92340824-92340824 on Build GRCh38
rs779688661 Chr.10: 92340785-92340785 on Build GRCh38

View Full Product Details