Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62813661-62813906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR2
Entrez Gene ID
1959
Gene Name
early growth response 2
Gene Aliases
AT591, CMT1D, CMT4E, KROX20
UniGene
Hs.1395
Gene Chromosome Location
Chr.10: 62811996-62819167 on Build GRCh38
Transcripts
NM_000399.4 NM_001136177.2 NM_001136178.1 NM_001136179.2 NM_001321037.1 XM_011539427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599061 c.772G>C p.V258L Chr.10: 62813866-62813866 on Build GRCh38
5837109 c.907G>A p.A303T Chr.10: 62813731-62813731 on Build GRCh38
3382948 c.896C>G p.S299* Chr.10: 62813742-62813742 on Build GRCh38
6036469 c.934C>G p.P312A Chr.10: 62813704-62813704 on Build GRCh38
684741 c.754C>A p.P252T Chr.10: 62813884-62813884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977162820 Chr.10: 62813714-62813714 on Build GRCh38
rs368282744 Chr.10: 62813869-62813869 on Build GRCh38
rs758709729 Chr.10: 62813864-62813864 on Build GRCh38
rs774265100 Chr.10: 62813775-62813775 on Build GRCh38
rs750955966 Chr.10: 62813861-62813861 on Build GRCh38
rs988041232 Chr.10: 62813818-62813818 on Build GRCh38
rs758589570 Chr.10: 62813837-62813837 on Build GRCh38
rs112474655 Chr.10: 62813729-62813729 on Build GRCh38
rs781325226 Chr.10: 62813885-62813885 on Build GRCh38
rs761862879 Chr.10: 62813808-62813808 on Build GRCh38
rs565202571 Chr.10: 62813781-62813781 on Build GRCh38
rs780441708 Chr.10: 62813868-62813868 on Build GRCh38
rs777368486 Chr.10: 62813727-62813729 on Build GRCh38
rs375890313 Chr.10: 62813795-62813795 on Build GRCh38
rs967096889 Chr.10: 62813709-62813709 on Build GRCh38
rs759852948 Chr.10: 62813776-62813776 on Build GRCh38
rs1044687586 Chr.10: 62813720-62813720 on Build GRCh38
rs759462455 Chr.10: 62813723-62813723 on Build GRCh38
rs775751575 Chr.10: 62813685-62813685 on Build GRCh38
rs763697987 Chr.10: 62813836-62813836 on Build GRCh38
rs754873661 Chr.10: 62813711-62813713 on Build GRCh38
rs908584883 Chr.10: 62813804-62813804 on Build GRCh38
rs772972643 Chr.10: 62813700-62813700 on Build GRCh38
rs765678040 Chr.10: 62813849-62813849 on Build GRCh38
rs886047093 Chr.10: 62813714-62813714 on Build GRCh38
rs766458625 Chr.10: 62813716-62813716 on Build GRCh38
rs777367425 Chr.10: 62813730-62813730 on Build GRCh38
rs753747037 Chr.10: 62813710-62813711 on Build GRCh38
rs752399953 Chr.10: 62813726-62813726 on Build GRCh38
rs768808272 Chr.10: 62813697-62813697 on Build GRCh38
rs774442561 Chr.10: 62813812-62813812 on Build GRCh38
rs565355765 Chr.10: 62813806-62813806 on Build GRCh38
rs532829227 Chr.10: 62813768-62813768 on Build GRCh38
rs746688326 Chr.10: 62813726-62813727 on Build GRCh38
rs104894158 Chr.10: 62813835-62813835 on Build GRCh38
rs759339694 Chr.10: 62813814-62813814 on Build GRCh38
rs774959041 Chr.10: 62813823-62813823 on Build GRCh38
rs764794586 Chr.10: 62813711-62813716 on Build GRCh38
rs772281054 Chr.10: 62813681-62813681 on Build GRCh38
rs746222548 Chr.10: 62813764-62813764 on Build GRCh38
rs747371531 Chr.10: 62813872-62813872 on Build GRCh38
rs760353922 Chr.10: 62813824-62813824 on Build GRCh38
rs760602079 Chr.10: 62813689-62813689 on Build GRCh38
rs758019499 Chr.10: 62813711-62813719 on Build GRCh38
rs863224359 Chr.10: 62813822-62813822 on Build GRCh38
rs200910043 Chr.10: 62813874-62813874 on Build GRCh38
rs563282095 Chr.10: 62813798-62813798 on Build GRCh38
rs1041555586 Chr.10: 62813839-62813839 on Build GRCh38
rs753339670 Chr.10: 62813741-62813741 on Build GRCh38
rs751393520 Chr.10: 62813717-62813717 on Build GRCh38
rs371616981 Chr.10: 62813761-62813761 on Build GRCh38
rs543533673 Chr.10: 62813759-62813759 on Build GRCh38
rs776440321 Chr.10: 62813866-62813866 on Build GRCh38
rs756821902 Chr.10: 62813838-62813838 on Build GRCh38
rs767452033 Chr.10: 62813819-62813819 on Build GRCh38
rs886047094 Chr.10: 62813720-62813720 on Build GRCh38

View Full Product Details