Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 15942201-15942468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYCN
Entrez Gene ID
4613
Gene Name
v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
Gene Aliases
MODED, N-myc, NMYC, ODED, bHLHe37
UniGene
Hs.25960
Gene Chromosome Location
Chr.2: 15940438-15947007 on Build GRCh38
Transcripts
NM_001293228.1 NM_001293231.1 NM_001293233.1 NM_005378.5 XM_017004168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
717282 c.333C>T p.I111I Chr.2: 15942397-15942397 on Build GRCh38
717284 c.225G>T p.P75P Chr.2: 15942289-15942289 on Build GRCh38
6956858 c.175delC p.P60fs*71 Chr.2: 15942239-15942239 on Build GRCh38
5899859 c.179C>T p.P60L Chr.2: 15942243-15942243 on Build GRCh38
1668725 c.170C>A p.P57H Chr.2: 15942234-15942234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771914958 Chr.2: 15942447-15942447 on Build GRCh38
rs573377492 Chr.2: 15942253-15942253 on Build GRCh38
rs750717149 Chr.2: 15942291-15942291 on Build GRCh38
rs780448923 Chr.2: 15942387-15942387 on Build GRCh38
rs754869267 Chr.2: 15942255-15942255 on Build GRCh38
rs886796492 Chr.2: 15942451-15942451 on Build GRCh38
rs996755231 Chr.2: 15942382-15942382 on Build GRCh38
rs761787091 Chr.2: 15942375-15942375 on Build GRCh38
rs751463329 Chr.2: 15942254-15942254 on Build GRCh38
rs773995882 Chr.2: 15942434-15942434 on Build GRCh38
rs780606114 Chr.2: 15942301-15942301 on Build GRCh38
rs764741267 Chr.2: 15942230-15942230 on Build GRCh38
rs747413221 Chr.2: 15942307-15942307 on Build GRCh38
rs748818541 Chr.2: 15942424-15942424 on Build GRCh38
rs769152073 Chr.2: 15942420-15942420 on Build GRCh38
rs541917011 Chr.2: 15942227-15942227 on Build GRCh38
rs911937395 Chr.2: 15942286-15942286 on Build GRCh38
rs772981756 Chr.2: 15942235-15942235 on Build GRCh38
rs777292565 Chr.2: 15942423-15942423 on Build GRCh38
rs748734356 Chr.2: 15942418-15942418 on Build GRCh38
rs758657333 Chr.2: 15942383-15942383 on Build GRCh38
rs865923398 Chr.2: 15942250-15942250 on Build GRCh38
rs767463815 Chr.2: 15942266-15942266 on Build GRCh38
rs751789215 Chr.2: 15942261-15942290 on Build GRCh38
rs759320806 Chr.2: 15942440-15942440 on Build GRCh38
rs769240539 Chr.2: 15942308-15942308 on Build GRCh38
rs1048338754 Chr.2: 15942439-15942439 on Build GRCh38
rs121913667 Chr.2: 15942295-15942295 on Build GRCh38
rs540489428 Chr.2: 15942438-15942438 on Build GRCh38
rs762639151 Chr.2: 15942245-15942245 on Build GRCh38
rs781535490 Chr.2: 15942403-15942403 on Build GRCh38
rs751918450 Chr.2: 15942397-15942397 on Build GRCh38
rs747547602 Chr.2: 15942319-15942319 on Build GRCh38
rs759339159 Chr.2: 15942356-15942356 on Build GRCh38
rs148975024 Chr.2: 15942310-15942310 on Build GRCh38
rs921199233 Chr.2: 15942392-15942392 on Build GRCh38
rs750579922 Chr.2: 15942380-15942380 on Build GRCh38
rs767373881 Chr.2: 15942359-15942359 on Build GRCh38
rs963929622 Chr.2: 15942364-15942364 on Build GRCh38
rs151113760 Chr.2: 15942343-15942343 on Build GRCh38
rs113994115 Chr.2: 15942281-15942281 on Build GRCh38
rs1052552775 Chr.2: 15942269-15942269 on Build GRCh38
rs41264199 Chr.2: 15942271-15942271 on Build GRCh38
rs779000620 Chr.2: 15942288-15942288 on Build GRCh38
rs774081987 Chr.2: 15942347-15942347 on Build GRCh38
rs769401203 Chr.2: 15942322-15942322 on Build GRCh38
rs770512842 Chr.2: 15942433-15942433 on Build GRCh38
rs757360369 Chr.2: 15942285-15942285 on Build GRCh38
rs755400695 Chr.2: 15942398-15942398 on Build GRCh38
rs752749374 Chr.2: 15942366-15942366 on Build GRCh38
rs772714029 Chr.2: 15942334-15942334 on Build GRCh38

View Full Product Details