Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185175857-185176102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP97
Entrez Gene ID
57587
Gene Name
cilia and flagella associated protein 97
Gene Aliases
KIAA1430, hmw
UniGene
Hs.535734
Gene Chromosome Location
Chr.4: 185159662-185210251 on Build GRCh38
Transcripts
NM_001292033.1 NM_020827.2 XM_017008483.1 XM_017008484.1 XM_017008485.1 XM_017008486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6694063 c.1096T>A p.F366I Chr.4: 185176010-185176010 on Build GRCh38
4695652 c.1191A>G p.E397E Chr.4: 185175915-185175915 on Build GRCh38
3130402 c.1077delA p.G360fs*12 Chr.4: 185176029-185176029 on Build GRCh38
1053751 c.1193T>C p.L398P Chr.4: 185175913-185175913 on Build GRCh38
5481881 c.1165C>T p.R389W Chr.4: 185175941-185175941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373907107 Chr.4: 185175960-185175960 on Build GRCh38
rs763355108 Chr.4: 185176047-185176047 on Build GRCh38
rs745592538 Chr.4: 185176028-185176029 on Build GRCh38
rs759778149 Chr.4: 185176067-185176068 on Build GRCh38
rs199637576 Chr.4: 185176012-185176012 on Build GRCh38
rs377061979 Chr.4: 185175963-185175963 on Build GRCh38
rs567292410 Chr.4: 185175976-185175976 on Build GRCh38
rs71593605 Chr.4: 185176065-185176066 on Build GRCh38
rs763192015 Chr.4: 185176066-185176066 on Build GRCh38
rs755645483 Chr.4: 185176075-185176075 on Build GRCh38
rs1023277788 Chr.4: 185175959-185175959 on Build GRCh38
rs773079777 Chr.4: 185175909-185175909 on Build GRCh38
rs1016371870 Chr.4: 185176045-185176045 on Build GRCh38
rs556874914 Chr.4: 185175967-185175967 on Build GRCh38
rs370176540 Chr.4: 185175978-185175978 on Build GRCh38
rs201895836 Chr.4: 185175890-185175890 on Build GRCh38
rs766770816 Chr.4: 185176057-185176057 on Build GRCh38
rs765465709 Chr.4: 185175947-185175947 on Build GRCh38
rs869140723 Chr.4: 185176076-185176077 on Build GRCh38
rs375689229 Chr.4: 185175987-185175987 on Build GRCh38
rs780714040 Chr.4: 185175971-185175971 on Build GRCh38
rs570390457 Chr.4: 185176036-185176036 on Build GRCh38
rs200291624 Chr.4: 185176065-185176065 on Build GRCh38
rs752254911 Chr.4: 185176074-185176074 on Build GRCh38
rs779476520 Chr.4: 185175889-185175889 on Build GRCh38
rs6855305 Chr.4: 185175891-185175891 on Build GRCh38
rs536319321 Chr.4: 185176068-185176068 on Build GRCh38
rs770234795 Chr.4: 185176062-185176063 on Build GRCh38
rs754542011 Chr.4: 185175894-185175894 on Build GRCh38
rs965059149 Chr.4: 185176072-185176072 on Build GRCh38
rs760820247 Chr.4: 185175940-185175940 on Build GRCh38
rs773654144 Chr.4: 185176044-185176044 on Build GRCh38
rs748157883 Chr.4: 185175904-185175904 on Build GRCh38
rs781085964 Chr.4: 185175901-185175901 on Build GRCh38
rs377617878 Chr.4: 185175948-185175948 on Build GRCh38
rs778399544 Chr.4: 185176011-185176012 on Build GRCh38
rs779474320 Chr.4: 185176057-185176058 on Build GRCh38
rs749104945 Chr.4: 185175986-185175986 on Build GRCh38
rs751174957 Chr.4: 185176062-185176062 on Build GRCh38
rs561532648 Chr.4: 185176060-185176062 on Build GRCh38
rs74754445 Chr.4: 185175945-185175945 on Build GRCh38
rs762163186 Chr.4: 185176028-185176028 on Build GRCh38
rs777242941 Chr.4: 185175944-185175944 on Build GRCh38
rs116235300 Chr.4: 185175957-185175957 on Build GRCh38
rs758760446 Chr.4: 185175955-185175955 on Build GRCh38
rs752526312 Chr.4: 185175973-185175973 on Build GRCh38
rs370181930 Chr.4: 185176075-185176076 on Build GRCh38
rs747272096 Chr.4: 185175993-185175993 on Build GRCh38
rs772343101 Chr.4: 185175915-185175915 on Build GRCh38
rs534539197 Chr.4: 185175922-185175922 on Build GRCh38
rs386402610 Chr.4: 185176072-185176073 on Build GRCh38
rs368666785 Chr.4: 185176006-185176006 on Build GRCh38
rs369749223 Chr.4: 185175941-185175941 on Build GRCh38
rs774067634 Chr.4: 185176044-185176054 on Build GRCh38
rs140276553 Chr.4: 185176076-185176076 on Build GRCh38
rs769438192 Chr.4: 185175907-185175907 on Build GRCh38
rs746018459 Chr.4: 185175913-185175913 on Build GRCh38
rs993431095 Chr.4: 185175899-185175899 on Build GRCh38
rs770333893 Chr.4: 185176037-185176037 on Build GRCh38

View Full Product Details