Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54190721-54190954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB5RL
Entrez Gene ID
606495
Gene Name
cytochrome b5 reductase like
Gene Aliases
-
UniGene
Hs.591426
Gene Chromosome Location
Chr.1: 54172336-54200073 on Build GRCh38
Transcripts
NM_001031672.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6507316 c.346C>T p.R116* Chr.1: 54190749-54190749 on Build GRCh38
5353055 c.218A>G p.N73S Chr.1: 54190877-54190877 on Build GRCh38
5353004 c.313C>T p.L105F Chr.1: 54190782-54190782 on Build GRCh38
2193669 c.228C>G p.T76T Chr.1: 54190867-54190867 on Build GRCh38
5016911 c.287G>A p.R96Q Chr.1: 54190808-54190808 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572197743 Chr.1: 54190815-54190815 on Build GRCh38
rs372078715 Chr.1: 54190822-54190822 on Build GRCh38
rs754974736 Chr.1: 54190919-54190919 on Build GRCh38
rs747427943 Chr.1: 54190935-54190935 on Build GRCh38
rs756215847 Chr.1: 54190749-54190749 on Build GRCh38
rs183984704 Chr.1: 54190773-54190773 on Build GRCh38
rs555399102 Chr.1: 54190803-54190803 on Build GRCh38
rs772275213 Chr.1: 54190847-54190847 on Build GRCh38
rs947907527 Chr.1: 54190890-54190890 on Build GRCh38
rs776224835 Chr.1: 54190776-54190776 on Build GRCh38
rs368704402 Chr.1: 54190809-54190809 on Build GRCh38
rs766756252 Chr.1: 54190899-54190899 on Build GRCh38
rs760967253 Chr.1: 54190898-54190898 on Build GRCh38
rs746127014 Chr.1: 54190748-54190748 on Build GRCh38
rs756413932 Chr.1: 54190820-54190820 on Build GRCh38
rs745457434 Chr.1: 54190854-54190854 on Build GRCh38
rs553055667 Chr.1: 54190914-54190914 on Build GRCh38
rs778105112 Chr.1: 54190934-54190934 on Build GRCh38
rs369271917 Chr.1: 54190879-54190879 on Build GRCh38
rs191854671 Chr.1: 54190767-54190767 on Build GRCh38
rs144642365 Chr.1: 54190929-54190929 on Build GRCh38
rs778123074 Chr.1: 54190851-54190851 on Build GRCh38
rs779330384 Chr.1: 54190838-54190838 on Build GRCh38
rs1031234084 Chr.1: 54190911-54190911 on Build GRCh38
rs769502665 Chr.1: 54190859-54190859 on Build GRCh38
rs750618240 Chr.1: 54190817-54190817 on Build GRCh38
rs557844186 Chr.1: 54190863-54190863 on Build GRCh38
rs976253788 Chr.1: 54190756-54190756 on Build GRCh38
rs140783073 Chr.1: 54190795-54190795 on Build GRCh38
rs759206614 Chr.1: 54190794-54190794 on Build GRCh38
rs763991187 Chr.1: 54190808-54190808 on Build GRCh38
rs541462144 Chr.1: 54190842-54190842 on Build GRCh38
rs1036973568 Chr.1: 54190792-54190792 on Build GRCh38
rs1030369016 Chr.1: 54190807-54190807 on Build GRCh38
rs374978053 Chr.1: 54190768-54190768 on Build GRCh38
rs371191047 Chr.1: 54190739-54190739 on Build GRCh38
rs61738954 Chr.1: 54190797-54190797 on Build GRCh38
rs762414782 Chr.1: 54190864-54190864 on Build GRCh38
rs755488923 Chr.1: 54190834-54190834 on Build GRCh38
rs924618170 Chr.1: 54190821-54190821 on Build GRCh38
rs781315078 Chr.1: 54190747-54190747 on Build GRCh38
rs780299398 Chr.1: 54190766-54190766 on Build GRCh38
rs374759352 Chr.1: 54190865-54190865 on Build GRCh38
rs895221636 Chr.1: 54190831-54190831 on Build GRCh38
rs371383333 Chr.1: 54190880-54190880 on Build GRCh38
rs770587611 Chr.1: 54190775-54190775 on Build GRCh38
rs767358017 Chr.1: 54190882-54190882 on Build GRCh38
rs990240283 Chr.1: 54190867-54190867 on Build GRCh38
rs774764544 Chr.1: 54190772-54190772 on Build GRCh38
rs376254778 Chr.1: 54190883-54190883 on Build GRCh38
rs758947711 Chr.1: 54190930-54190930 on Build GRCh38
rs922358840 Chr.1: 54190765-54190765 on Build GRCh38
rs779070727 Chr.1: 54190920-54190920 on Build GRCh38
rs749437839 Chr.1: 54190832-54190832 on Build GRCh38

View Full Product Details