Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13135522-13135783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NACC1
Entrez Gene ID
112939
Gene Name
nucleus accumbens associated 1
Gene Aliases
BEND8, BTBD14B, BTBD30, NAC-1, NAC1
UniGene
Hs.531614
Gene Chromosome Location
Chr.19: 13116848-13141147 on Build GRCh38
Transcripts
NM_052876.3 XM_005259721.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5485989 c.540C>T p.P180P Chr.19: 13135747-13135747 on Build GRCh38
7265712 c.431A>G p.Q144R Chr.19: 13135638-13135638 on Build GRCh38
3712804 c.324G>C p.Q108H Chr.19: 13135531-13135531 on Build GRCh38
4620733 c.377C>T p.P126L Chr.19: 13135584-13135584 on Build GRCh38
1258670 c.342G>A p.E114E Chr.19: 13135549-13135549 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754528785 Chr.19: 13135662-13135662 on Build GRCh38
rs777737473 Chr.19: 13135579-13135579 on Build GRCh38
rs781188008 Chr.19: 13135699-13135699 on Build GRCh38
rs751983812 Chr.19: 13135646-13135646 on Build GRCh38
rs774517762 Chr.19: 13135732-13135732 on Build GRCh38
rs770906465 Chr.19: 13135591-13135591 on Build GRCh38
rs374752584 Chr.19: 13135694-13135694 on Build GRCh38
rs769840684 Chr.19: 13135702-13135702 on Build GRCh38
rs778692787 Chr.19: 13135557-13135557 on Build GRCh38
rs776516112 Chr.19: 13135597-13135597 on Build GRCh38
rs868277418 Chr.19: 13135701-13135701 on Build GRCh38
rs142460106 Chr.19: 13135549-13135549 on Build GRCh38
rs767833951 Chr.19: 13135748-13135748 on Build GRCh38
rs866732640 Chr.19: 13135607-13135607 on Build GRCh38
rs947023395 Chr.19: 13135720-13135720 on Build GRCh38
rs1018544789 Chr.19: 13135609-13135609 on Build GRCh38
rs868294673 Chr.19: 13135764-13135764 on Build GRCh38
rs541005316 Chr.19: 13135725-13135725 on Build GRCh38
rs773750384 Chr.19: 13135753-13135753 on Build GRCh38
rs370514357 Chr.19: 13135627-13135627 on Build GRCh38
rs368599373 Chr.19: 13135558-13135558 on Build GRCh38
rs764392858 Chr.19: 13135645-13135645 on Build GRCh38
rs866163499 Chr.19: 13135667-13135667 on Build GRCh38
rs372693618 Chr.19: 13135651-13135651 on Build GRCh38
rs199695508 Chr.19: 13135751-13135751 on Build GRCh38
rs778617428 Chr.19: 13135671-13135671 on Build GRCh38
rs902911861 Chr.19: 13135726-13135726 on Build GRCh38
rs937184728 Chr.19: 13135698-13135698 on Build GRCh38
rs749471205 Chr.19: 13135711-13135711 on Build GRCh38
rs771489184 Chr.19: 13135585-13135585 on Build GRCh38
rs1009493804 Chr.19: 13135572-13135572 on Build GRCh38
rs141396248 Chr.19: 13135747-13135747 on Build GRCh38
rs1015067670 Chr.19: 13135608-13135608 on Build GRCh38
rs775567968 Chr.19: 13135710-13135710 on Build GRCh38
rs758160837 Chr.19: 13135684-13135684 on Build GRCh38
rs867964867 Chr.19: 13135733-13135733 on Build GRCh38
rs376241651 Chr.19: 13135659-13135659 on Build GRCh38
rs201757007 Chr.19: 13135688-13135688 on Build GRCh38
rs763209242 Chr.19: 13135630-13135630 on Build GRCh38
rs890857494 Chr.19: 13135570-13135570 on Build GRCh38
rs745831202 Chr.19: 13135700-13135700 on Build GRCh38
rs750947118 Chr.19: 13135660-13135660 on Build GRCh38
rs577024160 Chr.19: 13135693-13135693 on Build GRCh38
rs759650932 Chr.19: 13135611-13135611 on Build GRCh38
rs752352154 Chr.19: 13135683-13135683 on Build GRCh38
rs368756821 Chr.19: 13135763-13135763 on Build GRCh38
rs747447662 Chr.19: 13135543-13135543 on Build GRCh38
rs771973705 Chr.19: 13135578-13135578 on Build GRCh38
rs146491934 Chr.19: 13135612-13135612 on Build GRCh38
rs866968700 Chr.19: 13135745-13135745 on Build GRCh38
rs930075946 Chr.19: 13135685-13135685 on Build GRCh38

View Full Product Details