Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 30613678-30613935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRTAP6-1
Entrez Gene ID
337966
Gene Name
keratin associated protein 6-1
Gene Aliases
C21orf103, KAP6.1
UniGene
Hs.194301
Gene Chromosome Location
Chr.21: 30613686-30613904 on Build GRCh38
Transcripts
NM_181602.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5950165 c.183T>C p.Y61Y Chr.21: 30613722-30613722 on Build GRCh38
3405347 c.5G>A p.C2Y Chr.21: 30613900-30613900 on Build GRCh38
579010 c.147T>C p.Y49Y Chr.21: 30613758-30613758 on Build GRCh38
3405345 c.204C>A p.G68G Chr.21: 30613701-30613701 on Build GRCh38
6467872 c.131G>T p.R44I Chr.21: 30613774-30613774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200283642 Chr.21: 30613755-30613755 on Build GRCh38
rs755677753 Chr.21: 30613709-30613709 on Build GRCh38
rs770966738 Chr.21: 30613863-30613863 on Build GRCh38
rs372980382 Chr.21: 30613715-30613715 on Build GRCh38
rs772978984 Chr.21: 30613811-30613831 on Build GRCh38
rs778326655 Chr.21: 30613845-30613845 on Build GRCh38
rs776481848 Chr.21: 30613876-30613878 on Build GRCh38
rs748555459 Chr.21: 30613745-30613745 on Build GRCh38
rs757297747 Chr.21: 30613706-30613723 on Build GRCh38
rs755363811 Chr.21: 30613725-30613725 on Build GRCh38
rs1050773164 Chr.21: 30613726-30613726 on Build GRCh38
rs1041128260 Chr.21: 30613854-30613854 on Build GRCh38
rs750064287 Chr.21: 30613825-30613825 on Build GRCh38
rs144374144 Chr.21: 30613720-30613720 on Build GRCh38
rs114162209 Chr.21: 30613777-30613777 on Build GRCh38
rs772069308 Chr.21: 30613795-30613795 on Build GRCh38
rs113717308 Chr.21: 30613822-30613822 on Build GRCh38
rs74914596 Chr.21: 30613778-30613778 on Build GRCh38
rs200189534 Chr.21: 30613908-30613908 on Build GRCh38
rs375301612 Chr.21: 30613794-30613794 on Build GRCh38
rs1033323420 Chr.21: 30613768-30613768 on Build GRCh38
rs747839255 Chr.21: 30613780-30613781 on Build GRCh38
rs776679962 Chr.21: 30613901-30613901 on Build GRCh38
rs369092171 Chr.21: 30613886-30613886 on Build GRCh38
rs115002444 Chr.21: 30613762-30613762 on Build GRCh38
rs781443248 Chr.21: 30613720-30613749 on Build GRCh38
rs768400064 Chr.21: 30613747-30613748 on Build GRCh38
rs928667652 Chr.21: 30613838-30613838 on Build GRCh38
rs143074400 Chr.21: 30613887-30613887 on Build GRCh38
rs749415607 Chr.21: 30613852-30613852 on Build GRCh38
rs761444618 Chr.21: 30613902-30613902 on Build GRCh38
rs755395205 Chr.21: 30613830-30613830 on Build GRCh38
rs142155505 Chr.21: 30613716-30613716 on Build GRCh38
rs146113466 Chr.21: 30613900-30613900 on Build GRCh38
rs764794068 Chr.21: 30613906-30613906 on Build GRCh38
rs771199789 Chr.21: 30613765-30613765 on Build GRCh38
rs750117215 Chr.21: 30613907-30613907 on Build GRCh38
rs753459489 Chr.21: 30613807-30613807 on Build GRCh38
rs774199198 Chr.21: 30613781-30613786 on Build GRCh38
rs766047865 Chr.21: 30613911-30613911 on Build GRCh38
rs562505870 Chr.21: 30613823-30613823 on Build GRCh38
rs544198586 Chr.21: 30613800-30613800 on Build GRCh38
rs990663461 Chr.21: 30613846-30613846 on Build GRCh38
rs28567421 Chr.21: 30613744-30613744 on Build GRCh38
rs760298864 Chr.21: 30613810-30613811 on Build GRCh38
rs533036207 Chr.21: 30613831-30613831 on Build GRCh38
rs774586247 Chr.21: 30613869-30613869 on Build GRCh38
rs868166060 Chr.21: 30613828-30613828 on Build GRCh38
rs145876389 Chr.21: 30613795-30613795 on Build GRCh38
rs375647619 Chr.21: 30613753-30613753 on Build GRCh38
rs748691216 Chr.21: 30613835-30613835 on Build GRCh38
rs765962077 Chr.21: 30613833-30613833 on Build GRCh38
rs746078918 Chr.21: 30613719-30613720 on Build GRCh38
rs764091976 Chr.21: 30613801-30613801 on Build GRCh38
rs527457966 Chr.21: 30613897-30613897 on Build GRCh38
rs138494765 Chr.21: 30613844-30613844 on Build GRCh38
rs148838911 Chr.21: 30613759-30613759 on Build GRCh38
rs934964509 Chr.21: 30613913-30613913 on Build GRCh38
rs915177360 Chr.21: 30613791-30613791 on Build GRCh38
rs775216920 Chr.21: 30613888-30613888 on Build GRCh38

View Full Product Details