Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 36227077-36227310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNE
Entrez Gene ID
10020
Gene Name
glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase
Gene Aliases
DMRV, GLCNE, IBM2, NM, Uae1
UniGene
Hs.5920
Gene Chromosome Location
Chr.9: 36214441-36277056 on Build GRCh38
Transcripts
NM_001128227.2 NM_001190383.1 NM_001190384.1 NM_001190388.1 NM_005476.5 XM_005251334.4 XM_017014167.1 XM_017014168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4152297 c.1246G>A p.G416R Chr.9: 36227283-36227283 on Build GRCh38
1155125 c.1219G>A p.V407I Chr.9: 36227295-36227295 on Build GRCh38
4687457 c.1344G>A p.T448T Chr.9: 36227278-36227278 on Build GRCh38
4687460 c.1324G>T p.A442S Chr.9: 36227298-36227298 on Build GRCh38
1108900 c.1259G>A p.R420Q Chr.9: 36227270-36227270 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774260686 Chr.9: 36227252-36227252 on Build GRCh38
rs780092539 Chr.9: 36227270-36227270 on Build GRCh38
rs747199032 Chr.9: 36227271-36227271 on Build GRCh38
rs150174400 Chr.9: 36227188-36227189 on Build GRCh38
rs763953740 Chr.9: 36227233-36227233 on Build GRCh38
rs201025841 Chr.9: 36227242-36227242 on Build GRCh38
rs939258479 Chr.9: 36227189-36227189 on Build GRCh38
rs770532361 Chr.9: 36227202-36227202 on Build GRCh38
rs371881711 Chr.9: 36227287-36227287 on Build GRCh38
rs569986522 Chr.9: 36227131-36227131 on Build GRCh38
rs1057516364 Chr.9: 36227267-36227267 on Build GRCh38
rs16933102 Chr.9: 36227204-36227204 on Build GRCh38
rs866841518 Chr.9: 36227283-36227283 on Build GRCh38
rs542196416 Chr.9: 36227250-36227250 on Build GRCh38
rs144668576 Chr.9: 36227158-36227158 on Build GRCh38
rs904892113 Chr.9: 36227126-36227126 on Build GRCh38
rs115052162 Chr.9: 36227221-36227221 on Build GRCh38
rs185979147 Chr.9: 36227198-36227198 on Build GRCh38
rs759227946 Chr.9: 36227205-36227205 on Build GRCh38
rs750404513 Chr.9: 36227253-36227253 on Build GRCh38
rs1022557448 Chr.9: 36227235-36227235 on Build GRCh38
rs767017498 Chr.9: 36227214-36227214 on Build GRCh38
rs140784655 Chr.9: 36227284-36227284 on Build GRCh38
rs35638832 Chr.9: 36227262-36227262 on Build GRCh38
rs758086020 Chr.9: 36227278-36227278 on Build GRCh38
rs757251536 Chr.9: 36227247-36227247 on Build GRCh38
rs755383201 Chr.9: 36227274-36227274 on Build GRCh38
rs1048738084 Chr.9: 36227134-36227134 on Build GRCh38

View Full Product Details