Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1103991-1104238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX4
Entrez Gene ID
2879
Gene Name
glutathione peroxidase 4
Gene Aliases
GPx-4, GSHPx-4, MCSP, PHGPx, SMDS, snGPx, snPHGPx
UniGene
Hs.433951
Gene Chromosome Location
Chr.19: 1103926-1106789 on Build GRCh38
Transcripts
NM_001039847.2 NM_002085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4620590 c.22C>T p.R8C Chr.19: 1104065-1104065 on Build GRCh38
5695319 c.16C>T p.L6F Chr.19: 1104059-1104059 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs917166881 Chr.19: 1104040-1104040 on Build GRCh38
rs777613964 Chr.19: 1104026-1104026 on Build GRCh38
rs765996806 Chr.19: 1104092-1104092 on Build GRCh38
rs772251077 Chr.19: 1104053-1104053 on Build GRCh38
rs747381755 Chr.19: 1104065-1104065 on Build GRCh38
rs1023943971 Chr.19: 1104025-1104025 on Build GRCh38
rs1003429854 Chr.19: 1104180-1104180 on Build GRCh38
rs1046435919 Chr.19: 1104049-1104049 on Build GRCh38
rs976848690 Chr.19: 1104015-1104015 on Build GRCh38
rs529684277 Chr.19: 1104215-1104215 on Build GRCh38
rs8178967 Chr.19: 1104048-1104048 on Build GRCh38
rs577573367 Chr.19: 1104080-1104080 on Build GRCh38
rs769173951 Chr.19: 1104066-1104066 on Build GRCh38
rs905275808 Chr.19: 1104052-1104052 on Build GRCh38
rs72550710 Chr.19: 1104106-1104106 on Build GRCh38
rs573390223 Chr.19: 1104050-1104050 on Build GRCh38
rs925722714 Chr.19: 1104125-1104125 on Build GRCh38
rs955869469 Chr.19: 1104130-1104130 on Build GRCh38
rs1053017810 Chr.19: 1104038-1104038 on Build GRCh38
rs8178968 Chr.19: 1104152-1104152 on Build GRCh38
rs945018469 Chr.19: 1104078-1104078 on Build GRCh38
rs978503478 Chr.19: 1104119-1104119 on Build GRCh38
rs749330569 Chr.19: 1104033-1104033 on Build GRCh38
rs1026540214 Chr.19: 1104085-1104085 on Build GRCh38
rs757354426 Chr.19: 1104046-1104046 on Build GRCh38
rs544230793 Chr.19: 1104088-1104088 on Build GRCh38
rs912113378 Chr.19: 1104068-1104068 on Build GRCh38
rs1008526567 Chr.19: 1104090-1104090 on Build GRCh38
rs978555789 Chr.19: 1104024-1104024 on Build GRCh38
rs923701073 Chr.19: 1104028-1104028 on Build GRCh38
rs970697715 Chr.19: 1104034-1104034 on Build GRCh38
rs111442620 Chr.19: 1104158-1104158 on Build GRCh38
rs540807904 Chr.19: 1104059-1104059 on Build GRCh38
rs567653875 Chr.19: 1104094-1104094 on Build GRCh38
rs774076212 Chr.19: 1104104-1104104 on Build GRCh38
rs4807542 Chr.19: 1104079-1104079 on Build GRCh38
rs767130132 Chr.19: 1104107-1104107 on Build GRCh38
rs917133584 Chr.19: 1104172-1104172 on Build GRCh38
rs775567228 Chr.19: 1104087-1104087 on Build GRCh38
rs982101163 Chr.19: 1104218-1104218 on Build GRCh38

View Full Product Details