Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 50419547-50419772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMRTA2
Entrez Gene ID
63950
Gene Name
DMRT like family A2
Gene Aliases
-
UniGene
Hs.334481
Gene Chromosome Location
Chr.1: 50417550-50423447 on Build GRCh38
Transcripts
NM_032110.2 XM_011541937.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
358117 c.700C>T p.R234W Chr.1: 50419594-50419594 on Build GRCh38
1741944 c.710C>G p.S237* Chr.1: 50419584-50419584 on Build GRCh38
6063746 c.643A>C p.K215Q Chr.1: 50419651-50419651 on Build GRCh38
5753021 c.682A>C p.T228P Chr.1: 50419612-50419612 on Build GRCh38
5633519 c.636G>A p.P212P Chr.1: 50419658-50419658 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768712051 Chr.1: 50419707-50419707 on Build GRCh38
rs772872393 Chr.1: 50419581-50419581 on Build GRCh38
rs775445582 Chr.1: 50419683-50419683 on Build GRCh38
rs746793380 Chr.1: 50419588-50419588 on Build GRCh38
rs556550591 Chr.1: 50419568-50419568 on Build GRCh38
rs781012345 Chr.1: 50419668-50419668 on Build GRCh38
rs769754528 Chr.1: 50419676-50419676 on Build GRCh38
rs759525623 Chr.1: 50419594-50419594 on Build GRCh38
rs956460281 Chr.1: 50419662-50419662 on Build GRCh38
rs572985765 Chr.1: 50419754-50419754 on Build GRCh38
rs756679408 Chr.1: 50419570-50419570 on Build GRCh38
rs750798114 Chr.1: 50419625-50419625 on Build GRCh38
rs375713757 Chr.1: 50419648-50419648 on Build GRCh38
rs747637648 Chr.1: 50419634-50419634 on Build GRCh38
rs1005449713 Chr.1: 50419670-50419670 on Build GRCh38
rs776407763 Chr.1: 50419592-50419592 on Build GRCh38
rs1031796296 Chr.1: 50419657-50419657 on Build GRCh38
rs761076351 Chr.1: 50419746-50419746 on Build GRCh38
rs777295600 Chr.1: 50419653-50419653 on Build GRCh38
rs778617481 Chr.1: 50419571-50419571 on Build GRCh38
rs764241018 Chr.1: 50419608-50419608 on Build GRCh38
rs926590269 Chr.1: 50419744-50419744 on Build GRCh38
rs756728505 Chr.1: 50419631-50419631 on Build GRCh38
rs1278533 Chr.1: 50419646-50419646 on Build GRCh38
rs767820033 Chr.1: 50419738-50419738 on Build GRCh38
rs1036182518 Chr.1: 50419665-50419665 on Build GRCh38
rs61739961 Chr.1: 50419597-50419597 on Build GRCh38
rs750853133 Chr.1: 50419743-50419743 on Build GRCh38
rs765135618 Chr.1: 50419595-50419595 on Build GRCh38
rs575179236 Chr.1: 50419633-50419633 on Build GRCh38
rs768057755 Chr.1: 50419622-50419622 on Build GRCh38
rs762178055 Chr.1: 50419729-50419729 on Build GRCh38
rs745630474 Chr.1: 50419674-50419674 on Build GRCh38
rs377710327 Chr.1: 50419656-50419656 on Build GRCh38
rs967718232 Chr.1: 50419733-50419733 on Build GRCh38
rs774287820 Chr.1: 50419711-50419711 on Build GRCh38
rs747667218 Chr.1: 50419577-50419577 on Build GRCh38
rs775472647 Chr.1: 50419596-50419596 on Build GRCh38
rs751742435 Chr.1: 50419615-50419615 on Build GRCh38
rs762952261 Chr.1: 50419684-50419684 on Build GRCh38
rs371384511 Chr.1: 50419667-50419667 on Build GRCh38
rs770647861 Chr.1: 50419589-50419589 on Build GRCh38
rs757564673 Chr.1: 50419621-50419621 on Build GRCh38

View Full Product Details