Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40793140-40793365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT28
Entrez Gene ID
162605
Gene Name
keratin 28
Gene Aliases
K25IRS4, KRT25D
UniGene
Hs.59736
Gene Chromosome Location
Chr.17: 40792196-40799959 on Build GRCh38
Transcripts
NM_181535.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3517112 c.1239G>A p.G413G Chr.17: 40793168-40793168 on Build GRCh38
3771352 c.1197-9C>G p.? Chr.17: 40793219-40793219 on Build GRCh38
1610196 c.1223G>T p.G408V Chr.17: 40793184-40793184 on Build GRCh38
4983955 c.1247C>A p.S416Y Chr.17: 40793160-40793160 on Build GRCh38
6574480 c.1252+2T>C p.? Chr.17: 40793153-40793153 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868576420 Chr.17: 40793285-40793285 on Build GRCh38
rs761921022 Chr.17: 40793221-40793221 on Build GRCh38
rs992175482 Chr.17: 40793296-40793296 on Build GRCh38
rs774390408 Chr.17: 40793219-40793219 on Build GRCh38
rs565748465 Chr.17: 40793225-40793225 on Build GRCh38
rs536357888 Chr.17: 40793227-40793227 on Build GRCh38
rs777612768 Chr.17: 40793174-40793174 on Build GRCh38
rs770570549 Chr.17: 40793190-40793190 on Build GRCh38
rs542380881 Chr.17: 40793281-40793281 on Build GRCh38
rs746799851 Chr.17: 40793175-40793175 on Build GRCh38
rs986169777 Chr.17: 40793321-40793321 on Build GRCh38
rs999903579 Chr.17: 40793211-40793211 on Build GRCh38
rs758103839 Chr.17: 40793173-40793173 on Build GRCh38
rs769792221 Chr.17: 40793201-40793201 on Build GRCh38
rs537327011 Chr.17: 40793303-40793303 on Build GRCh38
rs960988662 Chr.17: 40793257-40793257 on Build GRCh38
rs745749466 Chr.17: 40793196-40793196 on Build GRCh38
rs760965869 Chr.17: 40793252-40793252 on Build GRCh38
rs570268631 Chr.17: 40793273-40793273 on Build GRCh38
rs142227683 Chr.17: 40793194-40793194 on Build GRCh38
rs1007032948 Chr.17: 40793178-40793178 on Build GRCh38
rs557475222 Chr.17: 40793306-40793306 on Build GRCh38
rs143597926 Chr.17: 40793256-40793256 on Build GRCh38
rs759915204 Chr.17: 40793262-40793262 on Build GRCh38
rs979508775 Chr.17: 40793312-40793312 on Build GRCh38
rs866992942 Chr.17: 40793192-40793192 on Build GRCh38

View Full Product Details