Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55809095-55809322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NLRP11
Entrez Gene ID
204801
Gene Name
NLR family pyrin domain containing 11
Gene Aliases
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7
UniGene
Hs.375039
Gene Chromosome Location
Chr.19: 55785397-55836762 on Build GRCh38
Transcripts
NM_001297743.1 NM_145007.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3539601 c.1403A>C p.N468T Chr.19: 55809207-55809207 on Build GRCh38
4127916 c.1317C>T p.S439S Chr.19: 55809293-55809293 on Build GRCh38
7276855 c.1291C>T p.Q431* Chr.19: 55809319-55809319 on Build GRCh38
1001672 c.1333C>T p.R445C Chr.19: 55809277-55809277 on Build GRCh38
7247631 c.1438G>A p.E480K Chr.19: 55809172-55809172 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746493018 Chr.19: 55809228-55809228 on Build GRCh38
rs775715079 Chr.19: 55809160-55809160 on Build GRCh38
rs763284926 Chr.19: 55809276-55809276 on Build GRCh38
rs147156673 Chr.19: 55809254-55809254 on Build GRCh38
rs780526258 Chr.19: 55809206-55809206 on Build GRCh38
rs12461110 Chr.19: 55809297-55809297 on Build GRCh38
rs757842969 Chr.19: 55809192-55809192 on Build GRCh38
rs751570988 Chr.19: 55809187-55809187 on Build GRCh38
rs568863144 Chr.19: 55809199-55809199 on Build GRCh38
rs769124769 Chr.19: 55809222-55809223 on Build GRCh38
rs757961174 Chr.19: 55809211-55809211 on Build GRCh38
rs776475161 Chr.19: 55809155-55809155 on Build GRCh38
rs770758532 Chr.19: 55809149-55809149 on Build GRCh38
rs144528374 Chr.19: 55809296-55809296 on Build GRCh38
rs764210174 Chr.19: 55809283-55809283 on Build GRCh38
rs866952822 Chr.19: 55809231-55809231 on Build GRCh38
rs1018547023 Chr.19: 55809146-55809146 on Build GRCh38
rs766227934 Chr.19: 55809245-55809245 on Build GRCh38
rs140090548 Chr.19: 55809263-55809263 on Build GRCh38
rs937380014 Chr.19: 55809217-55809217 on Build GRCh38
rs747013423 Chr.19: 55809132-55809132 on Build GRCh38
rs750900329 Chr.19: 55809200-55809200 on Build GRCh38
rs750997351 Chr.19: 55809291-55809291 on Build GRCh38
rs769611948 Chr.19: 55809256-55809256 on Build GRCh38
rs759333340 Chr.19: 55809158-55809158 on Build GRCh38
rs776564953 Chr.19: 55809236-55809236 on Build GRCh38
rs761805519 Chr.19: 55809289-55809289 on Build GRCh38
rs947216484 Chr.19: 55809245-55809247 on Build GRCh38
rs764284642 Chr.19: 55809284-55809284 on Build GRCh38
rs199615331 Chr.19: 55809253-55809253 on Build GRCh38
rs981428556 Chr.19: 55809184-55809184 on Build GRCh38
rs754238506 Chr.19: 55809300-55809300 on Build GRCh38
rs764275167 Chr.19: 55809182-55809182 on Build GRCh38
rs200402695 Chr.19: 55809215-55809215 on Build GRCh38
rs114531512 Chr.19: 55809205-55809205 on Build GRCh38
rs758133484 Chr.19: 55809303-55809303 on Build GRCh38
rs548284543 Chr.19: 55809208-55809208 on Build GRCh38
rs139431447 Chr.19: 55809159-55809159 on Build GRCh38
rs372266379 Chr.19: 55809288-55809288 on Build GRCh38
rs11880692 Chr.19: 55809233-55809233 on Build GRCh38
rs767654391 Chr.19: 55809290-55809290 on Build GRCh38

View Full Product Details