Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16361779-16362040 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPS15L1
Entrez Gene ID
58513
Gene Name
epidermal growth factor receptor pathway substrate 15 like 1
Gene Aliases
EPS15R
UniGene
Hs.654639
Gene Chromosome Location
Chr.19: 16355244-16472012 on Build GRCh38
Transcripts
NM_001258374.1 NM_001258375.1 NM_021235.2 NR_047665.1 NR_047666.1 XM_017027086.1 XM_017027087.1 XM_017027088.1 XM_017027089.1 XM_017027090.1 XM_017027091.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4947601 c.2458G>C p.D820H Chr.19: 16361907-16361907 on Build GRCh38
992440 c.2577C>T p.D859D Chr.19: 16361788-16361788 on Build GRCh38
74532 c.2458G>C p.D820H Chr.19: 16361907-16361907 on Build GRCh38
1196469 c.2426A>G p.E809G Chr.19: 16361939-16361939 on Build GRCh38
4909231 c.2533T>A p.S845T Chr.19: 16361832-16361832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752782676 Chr.19: 16361864-16361864 on Build GRCh38
rs148493117 Chr.19: 16361818-16361818 on Build GRCh38
rs761348291 Chr.19: 16361840-16361840 on Build GRCh38
rs141885889 Chr.19: 16362000-16362001 on Build GRCh38
rs144312610 Chr.19: 16361940-16361940 on Build GRCh38
rs756507720 Chr.19: 16361960-16361960 on Build GRCh38
rs767359234 Chr.19: 16361841-16361841 on Build GRCh38
rs868819541 Chr.19: 16361848-16361848 on Build GRCh38
rs756094810 Chr.19: 16362001-16362001 on Build GRCh38
rs747369495 Chr.19: 16361809-16361809 on Build GRCh38
rs750820714 Chr.19: 16361958-16361958 on Build GRCh38
rs551006641 Chr.19: 16361944-16361944 on Build GRCh38
rs766242583 Chr.19: 16361856-16361856 on Build GRCh38
rs542359984 Chr.19: 16361913-16361913 on Build GRCh38
rs146009121 Chr.19: 16361868-16361868 on Build GRCh38
rs759265656 Chr.19: 16361923-16361923 on Build GRCh38
rs377089622 Chr.19: 16361874-16361874 on Build GRCh38
rs755256417 Chr.19: 16361858-16361858 on Build GRCh38
rs561483634 Chr.19: 16361820-16361820 on Build GRCh38
rs757841102 Chr.19: 16361943-16361943 on Build GRCh38
rs747692461 Chr.19: 16361899-16361899 on Build GRCh38
rs994840907 Chr.19: 16361835-16361835 on Build GRCh38
rs541731564 Chr.19: 16361805-16361805 on Build GRCh38
rs751587680 Chr.19: 16361941-16361941 on Build GRCh38
rs747850262 Chr.19: 16361963-16361963 on Build GRCh38
rs752872450 Chr.19: 16361926-16361926 on Build GRCh38
rs1034975598 Chr.19: 16361870-16361870 on Build GRCh38
rs139911009 Chr.19: 16361914-16361914 on Build GRCh38
rs767374546 Chr.19: 16361831-16361833 on Build GRCh38
rs773159492 Chr.19: 16361845-16361845 on Build GRCh38
rs778168530 Chr.19: 16361872-16361872 on Build GRCh38
rs772572896 Chr.19: 16361912-16361912 on Build GRCh38
rs768419431 Chr.19: 16361831-16361831 on Build GRCh38
rs758957356 Chr.19: 16361802-16361802 on Build GRCh38
rs775238079 Chr.19: 16361998-16361998 on Build GRCh38
rs771590496 Chr.19: 16361970-16361970 on Build GRCh38
rs755224158 Chr.19: 16362001-16362002 on Build GRCh38
rs769628478 Chr.19: 16361995-16361995 on Build GRCh38
rs771560358 Chr.19: 16361903-16361903 on Build GRCh38
rs377387004 Chr.19: 16361988-16361988 on Build GRCh38
rs771280676 Chr.19: 16361817-16361817 on Build GRCh38
rs370996327 Chr.19: 16361883-16361883 on Build GRCh38
rs143071089 Chr.19: 16361801-16361801 on Build GRCh38
rs746484519 Chr.19: 16361972-16361972 on Build GRCh38
rs765477692 Chr.19: 16361924-16361924 on Build GRCh38
rs372459946 Chr.19: 16361851-16361851 on Build GRCh38
rs750421709 Chr.19: 16361999-16362000 on Build GRCh38
rs761716251 Chr.19: 16361802-16361803 on Build GRCh38
rs778727961 Chr.19: 16361961-16361961 on Build GRCh38
rs137911389 Chr.19: 16361836-16361836 on Build GRCh38
rs759353583 Chr.19: 16361992-16361992 on Build GRCh38
rs904993304 Chr.19: 16361839-16361839 on Build GRCh38
rs372467299 Chr.19: 16361971-16361971 on Build GRCh38
rs753543976 Chr.19: 16361857-16361857 on Build GRCh38
rs746221595 Chr.19: 16361893-16361893 on Build GRCh38
rs779202495 Chr.19: 16362009-16362011 on Build GRCh38
rs748219359 Chr.19: 16361800-16361800 on Build GRCh38
rs763065357 Chr.19: 16361935-16361935 on Build GRCh38
rs373976968 Chr.19: 16361979-16361979 on Build GRCh38
rs373537238 Chr.19: 16361891-16361891 on Build GRCh38
rs776463184 Chr.19: 16361920-16361920 on Build GRCh38
rs865997030 Chr.19: 16361868-16361868 on Build GRCh38
rs185020133 Chr.19: 16362001-16362001 on Build GRCh38
rs765428165 Chr.19: 16361863-16361863 on Build GRCh38
rs768437608 Chr.19: 16361898-16361898 on Build GRCh38

View Full Product Details