Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 99170328-99170589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MITD1
Entrez Gene ID
129531
Gene Name
microtubule interacting and trafficking domain containing 1
Gene Aliases
-
UniGene
Hs.14222
Gene Chromosome Location
Chr.2: 99161427-99181073 on Build GRCh38
Transcripts
NM_001320417.1 NM_001320418.1 NM_001320419.1 NM_138798.2 XM_011510581.2 XM_011510582.2 XM_011510583.1 XM_017003314.1 XM_017003315.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
722715 c.559C>G p.Q187E Chr.2: 99170571-99170571 on Build GRCh38
5863070 c.584delG p.R195fs*10 Chr.2: 99170546-99170546 on Build GRCh38
5001552 c.580G>T p.D194Y Chr.2: 99170550-99170550 on Build GRCh38
7107482 c.584G>A p.R195Q Chr.2: 99170546-99170546 on Build GRCh38
6712951 c.541G>A p.G181R Chr.2: 99170589-99170589 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773477322 Chr.2: 99170547-99170547 on Build GRCh38
rs374165760 Chr.2: 99170504-99170504 on Build GRCh38
rs911532050 Chr.2: 99170377-99170377 on Build GRCh38
rs748229588 Chr.2: 99170536-99170536 on Build GRCh38
rs147263719 Chr.2: 99170556-99170556 on Build GRCh38
rs946864812 Chr.2: 99170443-99170443 on Build GRCh38
rs949816277 Chr.2: 99170383-99170383 on Build GRCh38
rs770685103 Chr.2: 99170552-99170552 on Build GRCh38
rs935588919 Chr.2: 99170399-99170399 on Build GRCh38
rs749289729 Chr.2: 99170496-99170512 on Build GRCh38
rs570997838 Chr.2: 99170438-99170438 on Build GRCh38
rs144672860 Chr.2: 99170561-99170561 on Build GRCh38
rs991271998 Chr.2: 99170373-99170373 on Build GRCh38
rs141265262 Chr.2: 99170546-99170546 on Build GRCh38
rs981549342 Chr.2: 99170389-99170389 on Build GRCh38
rs562391698 Chr.2: 99170451-99170454 on Build GRCh38
rs915669555 Chr.2: 99170375-99170376 on Build GRCh38
rs376173468 Chr.2: 99170525-99170525 on Build GRCh38
rs779246508 Chr.2: 99170528-99170528 on Build GRCh38
rs1039856982 Chr.2: 99170469-99170469 on Build GRCh38
rs1052604238 Chr.2: 99170432-99170432 on Build GRCh38
rs950406473 Chr.2: 99170364-99170364 on Build GRCh38
rs909723202 Chr.2: 99170357-99170357 on Build GRCh38
rs749807457 Chr.2: 99170514-99170514 on Build GRCh38
rs962508589 Chr.2: 99170358-99170358 on Build GRCh38
rs532603448 Chr.2: 99170376-99170376 on Build GRCh38
rs966119315 Chr.2: 99170384-99170384 on Build GRCh38
rs924298101 Chr.2: 99170397-99170397 on Build GRCh38

View Full Product Details