Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1656207-1656466 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAMP1
Entrez Gene ID
57585
Gene Name
cramped chromatin regulator homolog 1
Gene Aliases
CRAMP1L, HN1L, TCE4
UniGene
Hs.603676
Gene Chromosome Location
Chr.16: 1614640-1677908 on Build GRCh38
Transcripts
NM_020825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3773741 c.1668C>T p.D556D Chr.16: 1656434-1656434 on Build GRCh38
4469497 c.1592C>T p.P531L Chr.16: 1656358-1656358 on Build GRCh38
4674569 c.1474G>A p.E492K Chr.16: 1656231-1656231 on Build GRCh38
4469480 c.1591C>T p.P531S Chr.16: 1656357-1656357 on Build GRCh38
6866698 c.1486G>T p.A496S Chr.16: 1656252-1656252 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370114636 Chr.16: 1656373-1656373 on Build GRCh38
rs749402671 Chr.16: 1656228-1656228 on Build GRCh38
rs770740244 Chr.16: 1656377-1656377 on Build GRCh38
rs373720836 Chr.16: 1656241-1656241 on Build GRCh38
rs61739873 Chr.16: 1656425-1656425 on Build GRCh38
rs772508584 Chr.16: 1656402-1656402 on Build GRCh38
rs754112928 Chr.16: 1656279-1656279 on Build GRCh38
rs199732432 Chr.16: 1656382-1656382 on Build GRCh38
rs766512234 Chr.16: 1656353-1656353 on Build GRCh38
rs145412892 Chr.16: 1656244-1656244 on Build GRCh38
rs758649118 Chr.16: 1656430-1656430 on Build GRCh38
rs186727389 Chr.16: 1656416-1656416 on Build GRCh38
rs921069720 Chr.16: 1656289-1656289 on Build GRCh38
rs201446092 Chr.16: 1656383-1656383 on Build GRCh38
rs1056706414 Chr.16: 1656351-1656351 on Build GRCh38
rs183769316 Chr.16: 1656363-1656363 on Build GRCh38
rs767706760 Chr.16: 1656345-1656345 on Build GRCh38
rs766016548 Chr.16: 1656242-1656242 on Build GRCh38
rs762158734 Chr.16: 1656341-1656341 on Build GRCh38
rs903117300 Chr.16: 1656295-1656295 on Build GRCh38
rs752981865 Chr.16: 1656399-1656399 on Build GRCh38
rs947261052 Chr.16: 1656292-1656292 on Build GRCh38
rs373328340 Chr.16: 1656375-1656375 on Build GRCh38
rs371446549 Chr.16: 1656364-1656364 on Build GRCh38
rs751887008 Chr.16: 1656255-1656255 on Build GRCh38
rs751286575 Chr.16: 1656440-1656440 on Build GRCh38
rs776911598 Chr.16: 1656417-1656417 on Build GRCh38
rs960409959 Chr.16: 1656266-1656266 on Build GRCh38
rs959779414 Chr.16: 1656227-1656227 on Build GRCh38
rs750534881 Chr.16: 1656275-1656275 on Build GRCh38
rs769502540 Chr.16: 1656395-1656395 on Build GRCh38
rs867399031 Chr.16: 1656360-1656360 on Build GRCh38
rs374860299 Chr.16: 1656419-1656419 on Build GRCh38
rs191664890 Chr.16: 1656236-1656236 on Build GRCh38
rs535159090 Chr.16: 1656401-1656401 on Build GRCh38
rs372185664 Chr.16: 1656270-1656270 on Build GRCh38
rs890199807 Chr.16: 1656378-1656378 on Build GRCh38
rs769410929 Chr.16: 1656333-1656333 on Build GRCh38
rs775959380 Chr.16: 1656312-1656312 on Build GRCh38
rs374738088 Chr.16: 1656370-1656370 on Build GRCh38
rs950653296 Chr.16: 1656420-1656420 on Build GRCh38
rs1026515397 Chr.16: 1656398-1656398 on Build GRCh38
rs748130394 Chr.16: 1656231-1656231 on Build GRCh38
rs764998478 Chr.16: 1656368-1656376 on Build GRCh38
rs764470515 Chr.16: 1656245-1656245 on Build GRCh38
rs552084892 Chr.16: 1656366-1656366 on Build GRCh38
rs995022425 Chr.16: 1656388-1656388 on Build GRCh38
rs747936661 Chr.16: 1656293-1656293 on Build GRCh38
rs562736193 Chr.16: 1656294-1656294 on Build GRCh38
rs577488354 Chr.16: 1656444-1656444 on Build GRCh38
rs754637328 Chr.16: 1656280-1656280 on Build GRCh38
rs201633864 Chr.16: 1656278-1656278 on Build GRCh38
rs948995216 Chr.16: 1656335-1656335 on Build GRCh38
rs950659348 Chr.16: 1656258-1656258 on Build GRCh38
rs759224794 Chr.16: 1656318-1656318 on Build GRCh38
rs377597921 Chr.16: 1656405-1656405 on Build GRCh38
rs778893305 Chr.16: 1656288-1656288 on Build GRCh38
rs772650332 Chr.16: 1656237-1656237 on Build GRCh38
rs540067077 Chr.16: 1656230-1656230 on Build GRCh38
rs746858691 Chr.16: 1656305-1656305 on Build GRCh38
rs774753825 Chr.16: 1656336-1656336 on Build GRCh38
rs200270407 Chr.16: 1656274-1656274 on Build GRCh38
rs770493494 Chr.16: 1656308-1656308 on Build GRCh38
rs757274453 Chr.16: 1656441-1656441 on Build GRCh38
rs777765671 Chr.16: 1656302-1656302 on Build GRCh38
rs374501457 Chr.16: 1656435-1656435 on Build GRCh38
rs915332314 Chr.16: 1656429-1656429 on Build GRCh38
rs759058105 Chr.16: 1656243-1656243 on Build GRCh38
rs780984929 Chr.16: 1656443-1656443 on Build GRCh38
rs773410575 Chr.16: 1656403-1656403 on Build GRCh38
rs750952543 Chr.16: 1656348-1656348 on Build GRCh38
rs537483186 Chr.16: 1656424-1656424 on Build GRCh38
rs761038864 Chr.16: 1656349-1656349 on Build GRCh38
rs61739874 Chr.16: 1656434-1656434 on Build GRCh38

View Full Product Details