Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 70175626-70175875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ2
Entrez Gene ID
3759
Gene Name
potassium voltage-gated channel subfamily J member 2
Gene Aliases
ATFB9, HHBIRK1, HHIRK1, IRK1, KIR2.1, LQT7, SQT3
UniGene
Hs.1547
Gene Chromosome Location
Chr.17: 70169535-70180044 on Build GRCh38
Transcripts
NM_000891.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6691158 c.601A>G p.I201V Chr.17: 70175640-70175640 on Build GRCh38
5240390 c.591C>T p.H197H Chr.17: 70175630-70175630 on Build GRCh38
3890284 c.801C>T p.I267I Chr.17: 70175840-70175840 on Build GRCh38
6147430 c.618C>A p.G206G Chr.17: 70175657-70175657 on Build GRCh38
707431 c.615C>G p.D205E Chr.17: 70175654-70175654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746781024 Chr.17: 70175749-70175749 on Build GRCh38
rs764523123 Chr.17: 70175676-70175676 on Build GRCh38
rs754883107 Chr.17: 70175765-70175765 on Build GRCh38
rs199473385 Chr.17: 70175818-70175818 on Build GRCh38
rs199473657 Chr.17: 70175718-70175718 on Build GRCh38
rs199473656 Chr.17: 70175689-70175689 on Build GRCh38
rs780976921 Chr.17: 70175768-70175768 on Build GRCh38
rs201850260 Chr.17: 70175714-70175714 on Build GRCh38
rs943311933 Chr.17: 70175756-70175756 on Build GRCh38
rs930806279 Chr.17: 70175844-70175844 on Build GRCh38
rs370571312 Chr.17: 70175656-70175656 on Build GRCh38
rs748150144 Chr.17: 70175775-70175775 on Build GRCh38
rs886039094 Chr.17: 70175789-70175789 on Build GRCh38
rs886042211 Chr.17: 70175845-70175845 on Build GRCh38
rs756826775 Chr.17: 70175703-70175703 on Build GRCh38
rs104894583 Chr.17: 70175685-70175685 on Build GRCh38
rs770964805 Chr.17: 70175783-70175783 on Build GRCh38
rs779856198 Chr.17: 70175733-70175733 on Build GRCh38
rs7221086 Chr.17: 70175699-70175699 on Build GRCh38
rs758144475 Chr.17: 70175723-70175723 on Build GRCh38
rs746052198 Chr.17: 70175838-70175838 on Build GRCh38
rs104894578 Chr.17: 70175691-70175691 on Build GRCh38
rs767844416 Chr.17: 70175654-70175654 on Build GRCh38
rs761179414 Chr.17: 70175660-70175660 on Build GRCh38
rs199473383 Chr.17: 70175683-70175683 on Build GRCh38
rs559935671 Chr.17: 70175717-70175717 on Build GRCh38
rs587781007 Chr.17: 70175812-70175812 on Build GRCh38
rs199473384 Chr.17: 70175692-70175692 on Build GRCh38
rs774461588 Chr.17: 70175820-70175820 on Build GRCh38
rs1037493754 Chr.17: 70175814-70175814 on Build GRCh38
rs138778849 Chr.17: 70175804-70175804 on Build GRCh38
rs141035459 Chr.17: 70175655-70175655 on Build GRCh38

View Full Product Details