Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 151136361-151136634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SEMA6C
Entrez Gene ID
10500
Gene Name
semaphorin 6C
Gene Aliases
SEMAY, m-SemaY, m-SemaY2
UniGene
Hs.516316
Gene Chromosome Location
Chr.1: 151131685-151148408 on Build GRCh38
Transcripts
NM_001178061.1 NM_001178062.1 NM_030913.4 XM_005244835.3 XM_017000075.1 XM_017000076.1 XM_017000077.1 XM_017000078.1 XM_017000079.1 XM_017000080.1 XM_017000081.1 XM_017000082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6459286 c.1027T>C p.F343L Chr.1: 151136527-151136527 on Build GRCh38
4995275 c.991G>A p.V331I Chr.1: 151136563-151136563 on Build GRCh38
6587187 c.1053G>T p.R351S Chr.1: 151136501-151136501 on Build GRCh38
6191947 c.995G>A p.C332Y Chr.1: 151136559-151136559 on Build GRCh38
4022128 c.1022G>A p.R341H Chr.1: 151136532-151136532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755179701 Chr.1: 151136456-151136456 on Build GRCh38
rs758194227 Chr.1: 151136608-151136608 on Build GRCh38
rs763055162 Chr.1: 151136405-151136406 on Build GRCh38
rs758967822 Chr.1: 151136425-151136425 on Build GRCh38
rs755234698 Chr.1: 151136409-151136409 on Build GRCh38
rs779940310 Chr.1: 151136431-151136431 on Build GRCh38
rs368620962 Chr.1: 151136546-151136546 on Build GRCh38
rs943309345 Chr.1: 151136571-151136571 on Build GRCh38
rs752068258 Chr.1: 151136558-151136558 on Build GRCh38
rs763302647 Chr.1: 151136551-151136551 on Build GRCh38
rs141765997 Chr.1: 151136517-151136517 on Build GRCh38
rs748433022 Chr.1: 151136468-151136468 on Build GRCh38
rs752077649 Chr.1: 151136407-151136407 on Build GRCh38
rs756424199 Chr.1: 151136583-151136583 on Build GRCh38
rs752924554 Chr.1: 151136570-151136570 on Build GRCh38
rs587640975 Chr.1: 151136509-151136509 on Build GRCh38
rs751668648 Chr.1: 151136439-151136439 on Build GRCh38
rs199848431 Chr.1: 151136404-151136404 on Build GRCh38
rs781554624 Chr.1: 151136458-151136458 on Build GRCh38
rs776241411 Chr.1: 151136531-151136531 on Build GRCh38
rs774795677 Chr.1: 151136500-151136500 on Build GRCh38
rs763740243 Chr.1: 151136406-151136406 on Build GRCh38
rs202109569 Chr.1: 151136413-151136413 on Build GRCh38
rs766908943 Chr.1: 151136556-151136556 on Build GRCh38
rs767992529 Chr.1: 151136425-151136425 on Build GRCh38
rs755089884 Chr.1: 151136564-151136564 on Build GRCh38
rs750856782 Chr.1: 151136419-151136419 on Build GRCh38
rs1021381604 Chr.1: 151136557-151136557 on Build GRCh38
rs745676304 Chr.1: 151136601-151136601 on Build GRCh38
rs370814864 Chr.1: 151136407-151136407 on Build GRCh38
rs753554673 Chr.1: 151136410-151136410 on Build GRCh38
rs778796779 Chr.1: 151136471-151136471 on Build GRCh38
rs765632732 Chr.1: 151136418-151136418 on Build GRCh38
rs200498855 Chr.1: 151136496-151136496 on Build GRCh38
rs375403121 Chr.1: 151136480-151136480 on Build GRCh38
rs767748385 Chr.1: 151136568-151136568 on Build GRCh38
rs761487418 Chr.1: 151136532-151136532 on Build GRCh38
rs778511882 Chr.1: 151136585-151136585 on Build GRCh38

View Full Product Details