Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 43566961-43567222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGK2
Entrez Gene ID
10110
Gene Name
SGK2, serine/threonine kinase 2
Gene Aliases
H-SGK2, dJ138B7.2
UniGene
Hs.300863
Gene Chromosome Location
Chr.20: 43558995-43585633 on Build GRCh38
Transcripts
NM_001199264.1 NM_016276.3 NM_170693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
723611 c.247G>T p.G83W Chr.20: 43567098-43567098 on Build GRCh38
7118697 c.254C>T p.S85L Chr.20: 43567105-43567105 on Build GRCh38
5824712 c.247G>A p.G83R Chr.20: 43567098-43567098 on Build GRCh38
4913390 c.247G>C p.G83R Chr.20: 43567098-43567098 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375329003 Chr.20: 43567085-43567085 on Build GRCh38
rs375815316 Chr.20: 43567023-43567023 on Build GRCh38
rs11554648 Chr.20: 43567070-43567070 on Build GRCh38
rs1052925652 Chr.20: 43567125-43567125 on Build GRCh38
rs922317944 Chr.20: 43567047-43567047 on Build GRCh38
rs200005007 Chr.20: 43567101-43567101 on Build GRCh38
rs780519103 Chr.20: 43567112-43567112 on Build GRCh38
rs1050011775 Chr.20: 43567093-43567093 on Build GRCh38
rs908205030 Chr.20: 43567014-43567014 on Build GRCh38
rs141029147 Chr.20: 43567114-43567114 on Build GRCh38
rs760032226 Chr.20: 43567164-43567164 on Build GRCh38
rs1032102806 Chr.20: 43567185-43567185 on Build GRCh38
rs749566189 Chr.20: 43567126-43567126 on Build GRCh38
rs773446020 Chr.20: 43567154-43567154 on Build GRCh38
rs769501682 Chr.20: 43567150-43567150 on Build GRCh38
rs1051809537 Chr.20: 43567081-43567081 on Build GRCh38
rs932521240 Chr.20: 43567060-43567060 on Build GRCh38
rs766388352 Chr.20: 43567159-43567159 on Build GRCh38
rs776539781 Chr.20: 43567126-43567126 on Build GRCh38
rs763091277 Chr.20: 43567157-43567157 on Build GRCh38
rs774471841 Chr.20: 43567160-43567160 on Build GRCh38
rs539478423 Chr.20: 43567134-43567134 on Build GRCh38
rs757882244 Chr.20: 43567044-43567044 on Build GRCh38
rs898083330 Chr.20: 43567051-43567051 on Build GRCh38
rs538000084 Chr.20: 43567007-43567007 on Build GRCh38
rs976539695 Chr.20: 43567035-43567035 on Build GRCh38
rs147630299 Chr.20: 43567000-43567000 on Build GRCh38
rs994114711 Chr.20: 43567053-43567053 on Build GRCh38
rs904471693 Chr.20: 43567100-43567100 on Build GRCh38
rs73620603 Chr.20: 43567025-43567025 on Build GRCh38

View Full Product Details