Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 105219918-105220179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL1RAPL2
Entrez Gene ID
26280
Gene Name
interleukin 1 receptor accessory protein like 2
Gene Aliases
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1
UniGene
Hs.675519
Gene Chromosome Location
Chr.X: 104566315-105769729 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

Gene Symbol
TEX13A
Entrez Gene ID
56157
Gene Name
testis expressed 13A
Gene Aliases
-
UniGene
Hs.567543
Gene Chromosome Location
Chr.X: 105218929-105220693 on Build GRCh38
Transcripts
NM_017416.1 XM_017029403.1 NM_001291277.1 NM_031274.4

SNP Detail

SNP ID Chromosome Location
rs782644244 Chr.X: 105220110-105220110 on Build GRCh38
rs41300159 Chr.X: 105219976-105219976 on Build GRCh38
rs782336037 Chr.X: 105219954-105219954 on Build GRCh38
rs782724760 Chr.X: 105220075-105220075 on Build GRCh38
rs782035203 Chr.X: 105220088-105220088 on Build GRCh38
rs199819714 Chr.X: 105220042-105220042 on Build GRCh38
rs782295077 Chr.X: 105220150-105220150 on Build GRCh38
rs782628378 Chr.X: 105220014-105220014 on Build GRCh38
rs782165401 Chr.X: 105219969-105219969 on Build GRCh38
rs191775065 Chr.X: 105219986-105219986 on Build GRCh38
rs781980945 Chr.X: 105219975-105219975 on Build GRCh38
rs782314162 Chr.X: 105220146-105220146 on Build GRCh38
rs782792744 Chr.X: 105219970-105219970 on Build GRCh38
rs782716716 Chr.X: 105219996-105219996 on Build GRCh38
rs782447109 Chr.X: 105220128-105220128 on Build GRCh38
rs782379050 Chr.X: 105219942-105219942 on Build GRCh38
rs150794186 Chr.X: 105219948-105219948 on Build GRCh38
rs782191085 Chr.X: 105220145-105220145 on Build GRCh38
rs371139667 Chr.X: 105219949-105219949 on Build GRCh38
rs782076281 Chr.X: 105219967-105219967 on Build GRCh38
rs781915856 Chr.X: 105220149-105220149 on Build GRCh38
rs782614639 Chr.X: 105220138-105220138 on Build GRCh38
rs372241672 Chr.X: 105220151-105220151 on Build GRCh38
rs782699507 Chr.X: 105220086-105220086 on Build GRCh38
rs183606927 Chr.X: 105219990-105219990 on Build GRCh38
rs782690962 Chr.X: 105220131-105220131 on Build GRCh38
rs781839599 Chr.X: 105220037-105220037 on Build GRCh38
rs782552729 Chr.X: 105220065-105220065 on Build GRCh38
rs781811593 Chr.X: 105220127-105220127 on Build GRCh38
rs782261289 Chr.X: 105220135-105220135 on Build GRCh38
rs781890931 Chr.X: 105220061-105220061 on Build GRCh38
rs782722878 Chr.X: 105219977-105219977 on Build GRCh38
rs781996269 Chr.X: 105220157-105220157 on Build GRCh38
rs373883548 Chr.X: 105220015-105220015 on Build GRCh38
rs782527369 Chr.X: 105220005-105220005 on Build GRCh38
rs782482739 Chr.X: 105220116-105220116 on Build GRCh38
rs782485677 Chr.X: 105220137-105220137 on Build GRCh38
rs782379757 Chr.X: 105220034-105220034 on Build GRCh38
rs781858444 Chr.X: 105220115-105220115 on Build GRCh38
rs782744693 Chr.X: 105220126-105220126 on Build GRCh38
rs782435399 Chr.X: 105219989-105219989 on Build GRCh38
rs782715384 Chr.X: 105220044-105220044 on Build GRCh38
rs913418680 Chr.X: 105220023-105220023 on Build GRCh38
rs782474353 Chr.X: 105220024-105220024 on Build GRCh38
rs782061168 Chr.X: 105220082-105220082 on Build GRCh38
rs782147727 Chr.X: 105220109-105220109 on Build GRCh38
rs992954982 Chr.X: 105220016-105220016 on Build GRCh38
rs781941580 Chr.X: 105220074-105220074 on Build GRCh38
rs782191180 Chr.X: 105220039-105220039 on Build GRCh38

View Full Product Details