Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 46651482-46651719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL10L
Entrez Gene ID
140801
Gene Name
ribosomal protein L10 like
Gene Aliases
RPL10_5_1358
UniGene
Hs.308332
Gene Chromosome Location
Chr.14: 46651013-46651825 on Build GRCh38
Transcripts
NM_080746.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5382282 c.233A>G p.K78R Chr.14: 46651504-46651504 on Build GRCh38
2197655 c.83A>G p.D28G Chr.14: 46651654-46651654 on Build GRCh38
6769158 c.236G>T p.S79I Chr.14: 46651501-46651501 on Build GRCh38
2197653 c.96C>T p.R32R Chr.14: 46651641-46651641 on Build GRCh38
2197649 c.205C>A p.R69S Chr.14: 46651532-46651532 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775298472 Chr.14: 46651663-46651663 on Build GRCh38
rs917574377 Chr.14: 46651562-46651562 on Build GRCh38
rs11557560 Chr.14: 46651612-46651612 on Build GRCh38
rs193920822 Chr.14: 46651643-46651643 on Build GRCh38
rs776689614 Chr.14: 46651572-46651572 on Build GRCh38
rs763538601 Chr.14: 46651695-46651695 on Build GRCh38
rs368319202 Chr.14: 46651689-46651689 on Build GRCh38
rs769703607 Chr.14: 46651521-46651521 on Build GRCh38
rs764517391 Chr.14: 46651587-46651588 on Build GRCh38
rs146301674 Chr.14: 46651685-46651685 on Build GRCh38
rs752024971 Chr.14: 46651593-46651593 on Build GRCh38
rs1051801955 Chr.14: 46651671-46651671 on Build GRCh38
rs763551550 Chr.14: 46651587-46651587 on Build GRCh38
rs747678911 Chr.14: 46651519-46651519 on Build GRCh38
rs949202562 Chr.14: 46651566-46651566 on Build GRCh38
rs200453552 Chr.14: 46651535-46651535 on Build GRCh38
rs980019816 Chr.14: 46651661-46651661 on Build GRCh38
rs773388593 Chr.14: 46651694-46651694 on Build GRCh38
rs776735096 Chr.14: 46651683-46651683 on Build GRCh38
rs770905515 Chr.14: 46651666-46651666 on Build GRCh38
rs138905696 Chr.14: 46651569-46651569 on Build GRCh38
rs780835176 Chr.14: 46651506-46651506 on Build GRCh38
rs141953926 Chr.14: 46651667-46651667 on Build GRCh38
rs754355435 Chr.14: 46651647-46651648 on Build GRCh38
rs374055419 Chr.14: 46651580-46651580 on Build GRCh38
rs780994512 Chr.14: 46651611-46651611 on Build GRCh38
rs370629655 Chr.14: 46651557-46651557 on Build GRCh38
rs201247193 Chr.14: 46651522-46651522 on Build GRCh38
rs960971618 Chr.14: 46651592-46651592 on Build GRCh38
rs768803836 Chr.14: 46651681-46651681 on Build GRCh38
rs150636660 Chr.14: 46651504-46651504 on Build GRCh38
rs760866228 Chr.14: 46651560-46651560 on Build GRCh38
rs562526716 Chr.14: 46651589-46651589 on Build GRCh38
rs367951246 Chr.14: 46651585-46651585 on Build GRCh38
rs969965076 Chr.14: 46651651-46651651 on Build GRCh38
rs1022379900 Chr.14: 46651563-46651563 on Build GRCh38
rs749063328 Chr.14: 46651555-46651555 on Build GRCh38
rs752193576 Chr.14: 46651627-46651627 on Build GRCh38
rs117580686 Chr.14: 46651608-46651608 on Build GRCh38
rs142878688 Chr.14: 46651578-46651578 on Build GRCh38
rs745732502 Chr.14: 46651670-46651670 on Build GRCh38
rs374478904 Chr.14: 46651635-46651635 on Build GRCh38
rs755652455 Chr.14: 46651574-46651577 on Build GRCh38
rs968227262 Chr.14: 46651568-46651568 on Build GRCh38
rs866727471 Chr.14: 46651642-46651642 on Build GRCh38
rs761909219 Chr.14: 46651687-46651687 on Build GRCh38

View Full Product Details