Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 146049024-146049283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3RF2
Entrez Gene ID
153769
Gene Name
SH3 domain containing ring finger 2
Gene Aliases
HEPP1, POSHER, PPP1R39, RNF158
UniGene
Hs.443728
Gene Chromosome Location
Chr.5: 145936563-146081773 on Build GRCh38
Transcripts
NM_152550.3 XM_005268380.2 XM_006714757.1 XM_011537567.2 XM_011537568.2 XM_011537569.2 XM_017009136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3784642 c.1162C>A p.L388M Chr.5: 146049085-146049085 on Build GRCh38
1723712 c.1211G>A p.G404E Chr.5: 146049134-146049134 on Build GRCh38
1720408 c.1225G>A p.V409I Chr.5: 146049148-146049148 on Build GRCh38
7111091 c.1280G>C p.R427P Chr.5: 146049203-146049203 on Build GRCh38
5618211 c.1265C>T p.S422F Chr.5: 146049188-146049188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776284711 Chr.5: 146049258-146049258 on Build GRCh38
rs367573674 Chr.5: 146049228-146049228 on Build GRCh38
rs374624666 Chr.5: 146049202-146049202 on Build GRCh38
rs201051050 Chr.5: 146049156-146049156 on Build GRCh38
rs548403479 Chr.5: 146049159-146049159 on Build GRCh38
rs561869294 Chr.5: 146049168-146049168 on Build GRCh38
rs770337153 Chr.5: 146049167-146049167 on Build GRCh38
rs371333971 Chr.5: 146049253-146049253 on Build GRCh38
rs745427899 Chr.5: 146049142-146049142 on Build GRCh38
rs747775775 Chr.5: 146049229-146049229 on Build GRCh38
rs148752800 Chr.5: 146049187-146049187 on Build GRCh38
rs200330959 Chr.5: 146049207-146049207 on Build GRCh38
rs139696891 Chr.5: 146049220-146049220 on Build GRCh38
rs753743799 Chr.5: 146049127-146049127 on Build GRCh38
rs780523237 Chr.5: 146049101-146049101 on Build GRCh38
rs377586859 Chr.5: 146049070-146049070 on Build GRCh38
rs1045662120 Chr.5: 146049256-146049256 on Build GRCh38
rs757995402 Chr.5: 146049200-146049200 on Build GRCh38
rs775982236 Chr.5: 146049059-146049059 on Build GRCh38
rs751401833 Chr.5: 146049178-146049178 on Build GRCh38
rs752488400 Chr.5: 146049115-146049115 on Build GRCh38
rs774594680 Chr.5: 146049236-146049236 on Build GRCh38
rs972206221 Chr.5: 146049182-146049182 on Build GRCh38
rs1052475790 Chr.5: 146049213-146049213 on Build GRCh38
rs146377978 Chr.5: 146049203-146049203 on Build GRCh38
rs139813276 Chr.5: 146049150-146049150 on Build GRCh38
rs767835486 Chr.5: 146049252-146049252 on Build GRCh38
rs750310157 Chr.5: 146049111-146049111 on Build GRCh38
rs910584957 Chr.5: 146049197-146049197 on Build GRCh38
rs36058593 Chr.5: 146049195-146049195 on Build GRCh38
rs777708342 Chr.5: 146049126-146049126 on Build GRCh38
rs749884545 Chr.5: 146049199-146049199 on Build GRCh38
rs749623952 Chr.5: 146049180-146049191 on Build GRCh38
rs755866307 Chr.5: 146049116-146049116 on Build GRCh38
rs769139489 Chr.5: 146049077-146049077 on Build GRCh38
rs759647712 Chr.5: 146049249-146049249 on Build GRCh38
rs765331245 Chr.5: 146049198-146049198 on Build GRCh38
rs144222014 Chr.5: 146049184-146049184 on Build GRCh38
rs1026680265 Chr.5: 146049120-146049120 on Build GRCh38
rs1055048523 Chr.5: 146049222-146049222 on Build GRCh38
rs373767672 Chr.5: 146049053-146049053 on Build GRCh38
rs763090331 Chr.5: 146049169-146049169 on Build GRCh38
rs761899238 Chr.5: 146049101-146049101 on Build GRCh38
rs548763256 Chr.5: 146049218-146049221 on Build GRCh38
rs763141840 Chr.5: 146049112-146049112 on Build GRCh38
rs770480202 Chr.5: 146049251-146049252 on Build GRCh38
rs138174522 Chr.5: 146049114-146049114 on Build GRCh38
rs545908874 Chr.5: 146049141-146049141 on Build GRCh38
rs776237219 Chr.5: 146049254-146049259 on Build GRCh38
rs753708354 Chr.5: 146049192-146049192 on Build GRCh38
rs563497572 Chr.5: 146049055-146049055 on Build GRCh38
rs370704589 Chr.5: 146049135-146049135 on Build GRCh38
rs770284250 Chr.5: 146049208-146049208 on Build GRCh38
rs35937996 Chr.5: 146049102-146049102 on Build GRCh38
rs747336583 Chr.5: 146049155-146049155 on Build GRCh38
rs34737526 Chr.5: 146049183-146049183 on Build GRCh38
rs529088702 Chr.5: 146049145-146049145 on Build GRCh38
rs750545470 Chr.5: 146049105-146049105 on Build GRCh38
rs748569627 Chr.5: 146049161-146049161 on Build GRCh38
rs769137101 Chr.5: 146049197-146049198 on Build GRCh38
rs375118719 Chr.5: 146049147-146049147 on Build GRCh38
rs777013577 Chr.5: 146049081-146049081 on Build GRCh38

View Full Product Details